Skip to Content
Merck
All Photos(2)

Documents

43732

Supelco

{2-Methyl-4-{{4-methyl-2-[4-(trifluoromethyl)phenyl]-5-thiazolyl}methylthio}phenoxy}acetic acid

analytical standard

Synonym(s):

Cardarine, Endurobol, GSK-516, GW 501516

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H18F3NO3S2
CAS Number:
Molecular Weight:
453.50
Beilstein:
18515150
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97.0% (CHN/S)
≥97.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

CC1=CC(SCC2=C(C)N=C(C3=CC=C(C(F)(F)F)C=C3)S2)=CC=C1OCC(O)=O

InChI

1S/C21H18F3NO3S2/c1-12-9-16(7-8-17(12)28-10-19(26)27)29-11-18-13(2)25-20(30-18)14-3-5-15(6-4-14)21(22,23)24/h3-9H,10-11H2,1-2H3,(H,26,27)

InChI key

YDBLKRPLXZNVNB-UHFFFAOYSA-N

General description

{2-Methyl-4-{{4-methyl-2-[4-(trifluoromethyl)phenyl]-5 thiazolyl}methylthio}phenoxy}acetic acid is a drug, which can act like a peroxisome proliferator-activated receptor agonist. It can play an important role in the regulation of central inflammation and can also be used to control brain inflammation process.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effects of the PPAR-? agonist GW501516 in an in vitro model of brain inflammation and antibody-induced demyelination
Defaux A, et al.
Journal of Neuroinflammation, 6, 15-15 (2009)
Antoinette Defaux et al.
Journal of neuroinflammation, 6, 15-15 (2009-05-09)
Brain inflammation plays a central role in numerous brain pathologies, including multiple sclerosis (MS). Microglial cells and astrocytes are the effector cells of neuroinflammation. They can be activated also by agents such as interferon-gamma (IFN-gamma) and lipopolysaccharide (LPS). Peroxisome proliferator-associated
Rémy Bosviel et al.
Free radical biology & medicine, 103, 146-154 (2016-12-19)
Whereas the anti-inflammatory properties and mechanisms of action of long chain ω3 PUFAs have been abundantly investigated, research gaps remain regarding the respective contribution and mechanisms of action of their oxygenated metabolites collectively known as oxylipins. We conducted a dose-dependent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service