Skip to Content
Merck
All Photos(1)

Documents

41244

Supelco

N-Desmethyl ofloxacin

analytical standard

Synonym(s):

9-Fluoro-2,3-dihydro-3-methyl-10-(1-piperazinyl)-7-oxo-7H-pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H18FN3O4
CAS Number:
Molecular Weight:
347.34
Beilstein:
4890029
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥96.5% (HPLC)

shelf life

limited shelf life, expiry date on the label

mp

256-262 °C

application(s)

clinical testing

format

neat

InChI

1S/C17H18FN3O4/c1-9-8-25-16-13-10(15(22)11(17(23)24)7-21(9)13)6-12(18)14(16)20-4-2-19-3-5-20/h6-7,9,19H,2-5,8H2,1H3,(H,23,24)

InChI key

WKRSSAPQZDHYRV-UHFFFAOYSA-N

General description

N-Desmethyl ofloxacin is one of the active metabolites of ofloxacin, a drug prescribed for treating bacterial infections (effective against both Gram-positive and Gram-negative bacteria) that acts by inhibiting the enzyme DNA gyrase.

Application

N-Desmethyl ofloxacin may be used as an analytical reference standard for the determination of the analyte in biological samples by high-pressure liquid chromatography with fluorescence detection and capillary electrophoresis using laser-induced fluorescence detection technique.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stereoselective determination of ofloxacin and its metabolites in human urine by capillary electrophoresis using laser-induced fluorescence detection
Horstkotter C and Blaschke G
Journal of Chromatography. B, Biomedical Sciences and Applications, 754(1), 169-178 (2001)
Kinetics of ofloxacin and its metabolites in cerebrospinal fluid after a single intravenous infusion of 400 milligrams of ofloxacin.
Nau R, et al.
Antimicrobial Agents and Chemotherapy, 38(8), 1849-1853 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service