Skip to Content
Merck
All Photos(1)

Documents

40200

Sigma-Aldrich

Dimethyldiphenylphosphonium iodide

purum, ≥98.0% (AT)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)2P(I)(CH3)2
CAS Number:
Molecular Weight:
342.16
Beilstein:
3637544
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

grade

purum

Assay

≥98.0% (AT)

form

solid

ign. residue

≤2.0%

mp

246-249 °C (lit.)
246-251 °C

SMILES string

[I-].C[P+](C)(c1ccccc1)c2ccccc2

InChI

1S/C14H16P.HI/c1-15(2,13-9-5-3-6-10-13)14-11-7-4-8-12-14;/h3-12H,1-2H3;1H/q+1;/p-1

InChI key

MGLUUYLPGOWCNW-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Other Notes

The corresponding di-ylide is a powerful reagent for the methylenation of carbonyl compounds; Ylide and di-ylide are used as ligands for organometallic compounds

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H. Schmidbaur et al.
Organometallics, 5, 1652-1652 (1986)
H. Schmidbaur
Accounts of Chemical Research, 8, 62-62 (1975)
H.J. Cristau et al.
Phosph. Sulfur Relat. Elem., 30, 135-135 (1987)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service