Skip to Content
Merck
All Photos(1)

Key Documents

36546

Supelco

Methyl chloroacetate

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2COOCH3
CAS Number:
Molecular Weight:
108.52
Beilstein:
506255
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

3.8 (vs air)

vapor pressure

10 mmHg ( 25 °C)
5.25 mmHg ( 20 °C)

product line

PESTANAL®

autoignition temp.

869 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

18.5 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.422 (lit.)

bp

130 °C/740 mmHg (lit.)

mp

−33 °C (lit.)

density

1.238 g/mL at 25 °C (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

COC(=O)CCl

InChI

1S/C3H5ClO2/c1-6-3(5)2-4/h2H2,1H3

InChI key

QABLOFMHHSOFRJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Methyl chloroacetate can find applications as a solvent in the liquid-liquid extraction of neutral compounds like alkylphenones, separated using micellar electrokinetic chromatography.[1]
Methyl chloroacetate can find applications as a solvent in the liquid-liquid extraction of neutral compounds like alkylphenones, separated using micellar electrokinetic chromatography.Methyl chloroacetate may be used as a precursor for the synthesis of d3-labeled dimethoate, an internal standard used for the quantification of dimethoate in food samples using isotope dilution and liquid chromatography coupled with electrospray ionization tandem mass spectrometry (LC/ESI-MS/MS) with multiple reaction monitoring mode (MRM).[2]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

116.6 °F - closed cup

Flash Point(C)

47 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Methyl chloroacetate as an extraction solvent for coupling liquid?liquid semimicroextraction with micellar electrokinetic chromatography through on-capillary decomposition for the separation of neutral compounds with concentration enhancement
Wang T, et al.
Journal of Chromatography A, 1147, 105-110 (2007)
Screening of dimethoate in food by isotope dilution and electrospray ionization tandem mass spectrometry
Mazzotti F, et al.
Rapid Communications in Mass Spectrometry, 23(10), 1515-1518 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service