Skip to Content
Merck
All Photos(1)

Documents

36408

Millipore

Bromcresol Purple Broth

NutriSelect® Plus, suitable for microbiology

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171606
NACRES:
NA.74

sterility

non-sterile

Quality Level

product line

BioChemika

form

powder

shelf life

limited shelf life, expiry date on the label

composition

beef extract, 3.0 g/L
bromo cresol purple, 0.04 g/L
peptic digest of animal tissue, 10.0 g/L
sodium chloride, 5.0 g/L

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbe id | utilization test: suitable
microbiological culture: suitable

final pH

7.0±0.2 (25 °C)

application(s)

food and beverages
food and beverages

microbiology

suitability

bacteria
nonselective and differential for Enterobacter spp.
nonselective and differential for Escherichia coli
nonselective and differential for Klebsiella spp.
nonselective and differential for Salmonella spp.
nonselective and differential for Shigella spp.
nonselective and differential for coliforms

Application

Bromcresol Purple Broth is recommended for studying fermentation of carbohydrates by pure culture

Preparation Note

Suspend 18 g in 1000 ml distilled water. Heat if necessary to dissolve the medium completely. Dispense in tubes containing inverted Durham`s tubes. Sterilize by autoclaving at 15 lbs pressure (121°C) for 10 minutes. Cool and aseptically add sterile desired carbohydrate to a final concentration of 0.5 - 1.0 %.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sharon Maes et al.
Journal of food protection, 82(2), 262-275 (2019-01-27)
After cleaning and disinfection (C&D), surface contamination can still be present in the production environment of food companies. Microbiological contamination on cleaned surfaces can be transferred to the manufactured food and consequently lead to foodborne illness and early food spoilage.

Articles

Salmonella, with 2,300+ serotypes, causes half of food-borne illnesses, often from dairy, poultry, and eggs.

Salmonella, with 2,300+ serotypes, causes half of food-borne illnesses, often from dairy, poultry, and eggs.

An article concerning selective growth media for differentiation and detection of Escherichia coli and other coliforms.

An article concerning selective growth media for differentiation and detection of Escherichia coli and other coliforms.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service