Skip to Content
Merck
All Photos(1)

Key Documents

34569

Supelco

Propachlor-(isopropyl-d7)

PESTANAL®, analytical standard

Synonym(s):

2-Chloro-N-isopropyl-d7-acetanilide, N-Isopropyl-d7-N-phenyl-chloroacetamide, Propachlor-d7

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11D7H7ClNO
Molecular Weight:
218.73
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

SMILES string

ClCC(N(C([2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])C1=CC=CC=C1)=O

InChI

1S/C11H14ClNO/c1-9(2)13(11(14)8-12)10-6-4-3-5-7-10/h3-7,9H,8H2,1-2H3/i1D3,2D3,9D

InChI key

MFOUDYKPLGXPGO-SCENNGIESA-N

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service