Skip to Content
Merck
All Photos(1)

Key Documents

34464

Supelco

Deterenol acetate

VETRANAL®, analytical standard

Synonym(s):

4-Hydroxy-α-[(isopropylamino)methyl]benzenemethanol acetate salt, N-Isopropyloctopamine acetate salt, PI 39 acetate salt, Paraproterenol acetate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H17NO2 · C2H4O2
CAS Number:
Molecular Weight:
255.31
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

application(s)

clinical testing

format

neat

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service