32995
Levothyroxine sodium hydrate
PESTANAL®, analytical standard
Synonym(s):
3,3′,5,5′-Tetraiodo-L-thyronine monosodium salt hydrate, L-Thyroxin monosodium salt hydrate, Sodium O-(4-hydroxy-3,5-diiodophenyl)-3,5-diiodo-L-tyrosine hydrate
About This Item
Recommended Products
grade
analytical standard
Quality Level
product line
PESTANAL®
shelf life
limited shelf life, expiry date on the label
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
agriculture
environmental
format
neat
SMILES string
O.[Na+].N[C@@H](Cc1cc(I)c(Oc2cc(I)c(O)c(I)c2)c(I)c1)C([O-])=O
InChI
1S/C15H11I4NO4.Na.H2O/c16-8-4-7(5-9(17)13(8)21)24-14-10(18)1-6(2-11(14)19)3-12(20)15(22)23;;/h1-2,4-5,12,21H,3,20H2,(H,22,23);;1H2/q;+1;/p-1/t12-;;/m0../s1
InChI key
ANMYAHDLKVNJJO-LTCKWSDVSA-M
Gene Information
human ... THRA(7067) , THRB(7068)
Looking for similar products? Visit Product Comparison Guide
Application
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
STOT RE 1
Target Organs
Thyroid,Cardio-vascular system,Kidney
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service