Skip to Content
Merck
All Photos(1)

Key Documents

32377

Supelco

erythro-Isoxsuprine-(phenoxy-13C6) hydrochloride

VETRANAL®, analytical standard

Synonym(s):

(1RS,2SR)-1-(4-Hydroxyphenyl)-2-[(1SR)1-methyl-2-phenoxy-13C6-ethylamino]propan-1-ol hydrochloride, (1RS,2SR,1′SR)-1-(p-Hydroxyphenyl)-2-(1′-methyl-2′-phenoxy-13C6-ethylamino)-1-propanol hydrochloride, mixture of enantiomers, erythro-Isoxsuprine-13C6 hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C6C12H23NO3 · HCl
Molecular Weight:
343.80
UNSPSC Code:
41116107

grade

analytical standard

product line

VETRANAL®

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service