Skip to Content
Merck
All Photos(2)

Key Documents

31681

Supelco

Thiometon in Shellsol

PESTANAL®, analytical standard

Synonym(s):

S-2-(Ethylthio)ethyl O,O′-dimethyl dithiophosphate solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Beilstein:
1707379
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture

format

neat

storage temp.

2-8°C

SMILES string

CCSCCSP(=S)(OC)OC

InChI

1S/C6H15O2PS3/c1-4-11-5-6-12-9(10,7-2)8-3/h4-6H2,1-3H3

InChI key

OPASCBHCTNRLRM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Thiometon is a systemic, organophosphorus insecticide and acaricide.[1][2] It is a cholinesterase inhibitor, used for controlling sucking insects in a wide variety of crops.[2]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Chronic 2 - Asp. Tox. 1 - Carc. 2 - Eye Irrit. 2

Supplementary Hazards

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Pesticide Encyclopedia (2014)
Toxicity of organophosphate insecticides and their metabolites to the water flea Daphnia magna, the Microtox test and an acetylcholinesterase inhibition test.
Galli R, et al.
Aquatic Toxicology (Amsterdam, Netherlands), 30(3), 259-269 (1994)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service