All Photos(3)
About This Item
Linear Formula:
C6H5CH=NNHSO2C6H4CH3
CAS Number:
Molecular Weight:
274.34
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47
Recommended Products
Assay
98%
form
powder, crystals or chunks
mp
130-131 °C (lit.)
application(s)
diagnostic assay manufacturing
hematology
histology
storage temp.
room temp
SMILES string
Cc1ccc(cc1)S(=O)(=O)N\N=C\c2ccccc2
InChI
1S/C14H14N2O2S/c1-12-7-9-14(10-8-12)19(17,18)16-15-11-13-5-3-2-4-6-13/h2-11,16H,1H3/b15-11+
InChI key
FZFLTDNAHASQQC-RVDMUPIBSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service