Skip to Content
Merck
All Photos(1)

Key Documents

23051

Sigma-Aldrich

4-(2-Chloroethyl)morpholine hydrochloride

purum, ≥97.0% (AT)

Synonym(s):

2-Morpholinoethyl chloride hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12ClNO · HCl
CAS Number:
Molecular Weight:
186.08
Beilstein:
3684083
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

purum

Quality Level

Assay

≥97.0% (AT)

form

solid

mp

180-185 °C (dec.) (lit.)
180-185 °C (dec.)

SMILES string

Cl[H].ClCCN1CCOCC1

InChI

1S/C6H12ClNO.ClH/c7-1-2-8-3-5-9-6-4-8;/h1-6H2;1H

InChI key

NBJHDLKSWUDGJG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service