Skip to Content
Merck
All Photos(1)

Key Documents

19734

Supelco

tert-Butylcalix[4]arene-tetrakis(N,N-dimethylacetamide)

Selectophore, ≥97.0%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C60H84N4O8
CAS Number:
Molecular Weight:
989.33
Beilstein:
6172613
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

product line

Selectophore

Quality Level

Assay

≥97.0% (TLC)
≥97.0%

form

solid

mp

≥240 °C

SMILES string

CN(C)C(=O)COc1c2Cc3cc(cc(Cc4cc(cc(Cc5cc(cc(Cc1cc(c2)C(C)(C)C)c5OCC(=O)N(C)C)C(C)(C)C)c4OCC(=O)N(C)C)C(C)(C)C)c3OCC(=O)N(C)C)C(C)(C)C

InChI

1S/C60H84N4O8/c1-57(2,3)45-25-37-21-39-27-46(58(4,5)6)29-41(54(39)70-34-50(66)62(15)16)23-43-31-48(60(10,11)12)32-44(56(43)72-36-52(68)64(19)20)24-42-30-47(59(7,8)9)28-40(55(42)71-35-51(67)63(17)18)22-38(26-45)53(37)69-33-49(65)61(13)14/h25-32H,21-24,33-36H2,1-20H3

InChI key

FTXOVFAXHOOZQB-UHFFFAOYSA-N

General description

Visit our Sensor Applications portal to learn more.

Application

tert-Butylcalix[4]arene-tetrakis(N,N-dimethylacetamide) may be used in the extraction of 14 lanthanoids along with thenoyltrifluoroacetone from perchlorate medium.

Other Notes

Starting material for the synthesis of ionophores, e.g. a lead ionophore

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P.L. Platten et al.
Journal of the American Chemical Society, 114, 10573-10573 (1992)
p-tert-Butylcalix [4] arene tetrakis (N, N-dimethylacetamide) as a second ligand in the complexation of trivalent lanthanoids with thenoyltrifluoroacetone in solution and investigation of a solid Eu (III) complex.
Atanassova, Maria, Nikolay Vassilev, and Ivan Dukov.
Separation Science and Technology, 78, 214-219 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service