Skip to Content
Merck
All Photos(1)

Documents

18858

Supelco

5-Aminosalicylic acid

analytical standard

Synonym(s):

5-AS, 5-Amino-2-hydroxybenzoic acid, Mesalamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H3-2-(OH)CO2H
CAS Number:
Molecular Weight:
153.14
Beilstein:
2090421
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.5% (NT)
≥99.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.5% water

mp

275-280 °C (dec.) (lit.)

application(s)

pharmaceutical

format

neat

storage temp.

2-8°C

SMILES string

Nc1ccc(O)c(c1)C(O)=O

InChI

1S/C7H7NO3/c8-4-1-2-6(9)5(3-4)7(10)11/h1-3,9H,8H2,(H,10,11)

InChI key

KBOPZPXVLCULAV-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
5-Aminosalicylic acid is a peroxidase substrate suitable for use in ELISA procedures. This substrate produces a soluble end product that is brown in color and can be read spectrophotometrically at 450 nm. The reaction may be stopped with 3 N NaOH and read at 550 nm.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rosario Cuomo et al.
United European gastroenterology journal, 2(5), 413-442 (2014-11-02)
The statements produced by the Consensus Conference on Diverticular Disease promoted by GRIMAD (Gruppo Italiano Malattia Diverticolare, Italian Group on Diverticular Diseases) are reported. Topics such as epidemiology, risk factors, diagnosis, medical and surgical treatment of diverticular disease (DD) in
Grzegorz Garbacz et al.
The Journal of pharmacy and pharmacology, 67(2), 199-208 (2015-01-06)
For the treatment of inflammatory bowel disease, the development of pH responsive modified release dosage forms is one of the most common approaches to achieve targeted drug delivery. In this study, the dissolution behaviour of eight different modified release (MR)
Alan C Moss et al.
The American journal of gastroenterology, 109(12), 1850-1855 (2014-06-11)
Poor adherence to mesalamine is common and driven by a combination of lifestyle and behavioral factors, as well as health beliefs. We sought to develop a valid tool to identify barriers to patient adherence and predict those at risk for
Natalia Pedersen et al.
Inflammatory bowel diseases, 20(12), 2276-2285 (2014-09-24)
To individualize treatment with mesalazine for ulcerative colitis relapses through a self-managed, web-based solution to optimize the short-term disease course. Prospective, open-label, web-guided study with 3 months mesalazine therapy among patients with mild-to-moderate ulcerative colitis. Once a week, patients completed
W J Sandborn et al.
Alimentary pharmacology & therapeutics, 41(5), 409-418 (2015-01-16)
Conventional oral corticosteroids are effective at reducing inflammation associated with ulcerative colitis (UC); however, systemic adverse effects limit their use. Budesonide MMX is an extended-release, second-generation corticosteroid that targets delivery of budesonide to the entire colon. To analyse efficacy and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service