Skip to Content
Merck
All Photos(1)

Key Documents

37-0060

Sigma-Aldrich

Carbol-Fuchsin solution according to Ziehl-Neelsen

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
Pricing and availability is not currently available.

form

liquid

availability

available only in Japan

SMILES string

Cl[H].Cc1cc(ccc1N)\C(=C2\C=CC(=N)C=C2)c3ccc(N)c(C)c3

InChI

1S/C21H21N3.ClH/c1-13-11-16(5-9-19(13)23)21(15-3-7-18(22)8-4-15)17-6-10-20(24)14(2)12-17;/h3-12,22H,23-24H2,1-2H3;1H

InChI key

YCIVZWKFFBDLTG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Suitability

nuclear and bacterial staining

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Muta. 2 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service