Skip to Content
Merck
All Photos(1)

Key Documents

28-6120

Sigma-Aldrich

Sulfur monochloride

CP

Synonym(s):

Sulfur chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
S2Cl2
CAS Number:
Molecular Weight:
135.04
EC Number:
MDL number:
UNSPSC Code:
12352307
PubChem Substance ID:
grade:
CP
form:
liquid
Pricing and availability is not currently available.

grade

CP

vapor density

4.7 (vs air)

vapor pressure

6.8 mmHg ( 20 °C)

form

liquid

autoignition temp.

451 °F

availability

available only in Japan

refractive index

n20/D 1.658 (lit.)

bp

138 °C (lit.)

mp

−80 °C (lit.)

density

1.688 g/mL at 25 °C (lit.)

SMILES string

ClSSCl

InChI

1S/Cl2S2/c1-3-4-2

InChI key

PXJJSXABGXMUSU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Skin Corr. 1A

Supplementary Hazards

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel K W Mok et al.
The Journal of chemical physics, 125(10), 104303-104303 (2006-09-27)
Geometry optimization calculations were carried out on the (approximate)X (1)A(1) state of SCl(2) and the (approximate)X(2)B(1), (approximate)A(2)B(2), (approximate)B(2)A(1), (approximate)C(2)A(1), (approximate)D(2)A(2), and (approximate)E (2)B(2) states of SCl(2) (+) at the restricted-spin coupled-cluster single-double plus perturbative triple excitation [RCCSD(T)] level with basis
[Eye burn caused by sulfur monochloride].
J Peyresblanques et al.
Bulletin des societes d'ophtalmologie de France, 87(3), 429-430 (1987-03-01)
M J Davies et al.
Free radical research, 33(6), 719-729 (2001-03-10)
Activated phagocytic cells generate hypochlorite (HOCl) via release of hydrogen peroxide and the enzyme myeloperoxidase. HOCl plays an important role in bacterial cell killing, but excessive or misplaced production of HOCI is also known to cause tissue damage. Studies have
F Machetti et al.
Advances in experimental medicine and biology, 483, 399-401 (2002-01-15)
(+/-)trans 2-Aminocyclohexanesulfonic acid and (+/-)trans 2-aminocyclopentanesulfonic acid were prepared from cyclohexene and cyclopentene respectively by sulfur monochloride addition, followed by oxidation to 2-chlorosulfonic acid and substitution of chlorine.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service