Skip to Content
Merck
All Photos(1)

Key Documents

05-4620

Sigma-Aldrich

Chromium(VI) oxide

CP

Synonym(s):

Chromic anhydride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CrO3
CAS Number:
Molecular Weight:
99.99
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

CP

availability

available only in Japan

mp

196 °C (dec.) (lit.)

SMILES string

O=[Cr](=O)=O

InChI

1S/Cr.3O

InChI key

WGLPBDUCMAPZCE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A - Eye Dam. 1 - Muta. 1B - Ox. Sol. 1 - Repr. 2 - Resp. Sens. 1 - Skin Corr. 1A - Skin Sens. 1 - STOT RE 1 Inhalation - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

EU REACH Annex XIV (Authorisation List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sethsiri S Samaratunga et al.
Environmental science and pollution research international, 15(1), 27-30 (2008-03-01)
Chromium enters into the aquatic environment as a result of effluent discharge from steel works, electroplating, leather tanning industries and chemical industries. As the Cr(VI) is very harmful to living organisms, it should be quickly removed from the environment when
Kyung-Hwa Son et al.
Journal of toxicology and environmental health. Part A, 67(13), 1027-1049 (2004-06-19)
CrO3 is cytotoxic for human epithelial 293 kidney cells over a narrow concentration range of approximately 2-8 microM (D50 approximately 3.0 microM); significantly greater toxicity is observed in clonogenic assays (D50 approximately 0.1-1.0 microM). Survival of a small fraction of
Sara L Holland et al.
Toxicology in vitro : an international journal published in association with BIBRA, 24(6), 1764-1767 (2010-07-20)
Toxicity of the environmental carcinogen chromate is known to involve sulfur starvation and also error-prone mRNA translation. Here we reconcile those facts using the yeast model. We demonstrate that: (i) cysteine and methionine starvation mimic Cr-induced translation errors, (ii) genetic
Evidence for the protective effect of ascorbic acid (vitamin C) in treatment with gamma-rays and chromium (VI) oxide (CrO3) in somatic cells of Drosophila.
O Olvera et al.
Mutation research, 346(1), 19-21 (1995-01-01)
Reyes Sierra-Alvarez
Waste management (New York, N.Y.), 29(6), 1885-1891 (2009-01-13)
Large volumes of preservative-treated wood containing toxic Cr, Cu and As salts are decommissioned worldwide. This study investigated the effectiveness of solid-state fermentation with copper-tolerant brown-rot fungi for the remediation of wood treated with chromated copper arsenate (CCA) and acid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service