Skip to Content
Merck
All Photos(1)

Documents

RIE156324272

Rieke Metals, Inc.

cyclopropyl 4-(morpholinomethyl)phenyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
245.32
MDL number:

Iupac Name

cyclopropyl[4-(4-morpholinylmethyl)phenyl]methanone

Inchi Code

1S/C15H19NO2/c17-15(14-5-6-14)13-3-1-12(2-4-13)11-16-7-9-18-10-8-16/h1-4,14H,5-11H2

InChI key

IUWLNNWDEVXWPK-UHFFFAOYSA-N

Purity

97%

Country of Origin

US

Product Link

https://riekemetals.com/products-services/catnum/6124-0101-00

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service