ZFTPS42TC
Level sensor
For use with Milli-Q® HX and HR 7000 Water Purification Systems
Synonym(s):
Level sensor 35X HT 4-20mA
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
packaging
pkg of 1 unit
parameter
pressure range at 1 bar (30 bar)
temp. range at -20 to 150 °C (storage)
compatibility
for use with Milli-Q® HR
for use with Milli-Q® HX
Looking for similar products? Visit Product Comparison Guide
General description
These piezoresistive transmitters are specially designed for applications in bioreactors and autoclaves. The transmitters can be used up to +150 °C, a temperature at which they can work continuously.
The sensor element is a chip of silicon, micro-machined. An independent temperature sensor is associated with the pressure measuring element.
The sensor element is a chip of silicon, micro-machined. An independent temperature sensor is associated with the pressure measuring element.
Legal Information
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service