Skip to Content
Merck
All Photos(1)

Key Documents

TANKLK001

Milli-Q

Water sensor

For use with RiOs and Elix® water purification systems

Synonym(s):

Water Sensor with Cable

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€172.00

€172.00


Estimated to ship on01 July 2025Details


Request more information

Select a Size

Change View
1 EA
€172.00

About This Item

UNSPSC Code:
41104200
eCl@ss:
33050190
NACRES:
JA.14

€172.00


Estimated to ship on01 July 2025Details


Request more information

feature

Sense water, shut down the system and closes the inlet solenoid valve.

packaging

pkg of 1 unit

cable L

4 m

compatibility

for use with Elix®
for use with RiOs

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

General description

The water sensor accessory for RiOs and Elix® systems detect the presence of water and automatically shut down the feedwater supply to the system. Thereby protecting the lab from water damage by isolating the system from the mainline if a leak occurs.

The sensors are usually placed on the floor or benchtop, where it is more convenient for the lab.

Application

This water sensor accessory for RiOs 30/50/100/150/200 and Elix® 20/35/70/100 water purification systems prevent water damage to the lab, in an unlikely event of a leak.

Features and Benefits

  • Reduces the risk of flooding: no damage to the lab or other instruments.
  • Cable length: 4 meters.

Legal Information

ELIX is a registered trademark of Merck KGaA, Darmstadt, Germany
RIOS is a trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service