Skip to Content
Merck
All Photos(1)

Key Documents

SX0785

Supelco

Sodium sulfite

anhydrous, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2SO3
CAS Number:
Molecular Weight:
126.04
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-821-4
Assay:
≥98.0% (iodometric)
form:
powder
solubility:
220 g/L
To inquire about this product SX0785, please contact your local Merck office or dealer. Contact Technical Service

Quality Level

Assay

≥98.0% (iodometric)

form

powder

impurities

≤0.005% Insoluble matter
≤0.03 meq/g Titrable free base

pH

8.8-10 (20 °C, 50 g/L in H2O)

mp

>500 °C (decomposition)

solubility

220 g/L

anion traces

chloride (Cl-): ≤0.02%

cation traces

Fe: ≤0.001%
heavy metals (as Pb): ≤0.001%

shipped in

ambient

storage temp.

room temp

SMILES string

[Na+].[Na+].[O-]S([O-])=O

InChI

1S/2Na.H2O3S/c;;1-4(2)3/h;;(H2,1,2,3)/q2*+1;/p-2

InChI key

GEHJYWRUCIMESM-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application


  • Comparative Studies of Organic Contaminant Removal in Different Calcium Sulfite-Enhanced Oxidant/Fe(II) Systems: This research explores the effectiveness of sodium sulfite in enhancing the removal of organic contaminants through various oxidant systems. (Zhou et al., 2023).

  • Review of Traditional and Emerging Residual Chlorine Quenchers on Disinfection By-Products: Focuses on the role of sodium sulfite as a residual chlorine quencher in water treatment processes, analyzing its impact on reducing harmful by-products and enhancing water quality (Li et al., 2023).

  • Boosting the Photoelectrochemical Performance of Bismuth Vanadate Photoanode Through Homojunction Construction: Investigates how sodium sulfite can improve the photoelectrochemical properties of bismuth vanadate, significant for renewable energy applications, particularly in solar energy conversion (Wang et al., 2023).

  • Light-driven Oxidative Demethylation Reaction Catalyzed by a Rieske-type Non-heme Iron Enzyme Stc2: Demonstrates the use of sodium sulfite in enzymatic reactions to achieve oxidative demethylation, which is crucial for synthetic biology and enzyme engineering (Hu et al., 2022).

  • Preparation of Nano Disperse Dyes Using Sulfomethylated Lignin: Effects of Sulfonic Group Contents: Highlights the application of sodium sulfite in the preparation of environmentally friendly nano disperse dyes from lignin, emphasizing its potential in sustainable material science (Tang et al., 2023).

Analysis Note

Assay (iodometric): 98.0% min
Chloride (Cl): 0.02% max
Free acid: To pass test
Heavy metals (as Pb): 0.001% max
Insoluble matter: 0.005% max
Iron (Fe): 0.001% max
Titrable free base: 0.03meq/g max

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service