Skip to Content
Merck
All Photos(2)

Key Documents

SCT029

Sigma-Aldrich

BioTracker 529 Green Pluripotent Stem Cell Dye

Live cell imaging dye for pluripotent stem cells that selectively identifies undifferentiated human ES and iPS cells.

Synonym(s):

Live cell imaging probe

Sign Into View Organizational & Contract Pricing

Select a Size

5 X 10 μG
€712.00

€712.00


Please contact Customer Service for Availability


Select a Size

Change View
5 X 10 μG
€712.00

About This Item

UNSPSC Code:
12352207
eCl@ss:
32161000
NACRES:
NA.47

€712.00


Please contact Customer Service for Availability

technique(s)

cell based assay: suitable

detection method

fluorometric

shipped in

ambient

General description

Pluripotent stem cells, including embryonic stem cells (ES cells) and induced pluripotent stem cells (iPSCs), have the capacity to give rise to differentiated progeny representative of all three germ layers (ectoderm, endoderm, and mesoderm). Characterization of human iPS cells includes alkaline phosphatase and antibody staining for Oct-4, Sox-2, Nanog, SSEA and TRA surface markers. These assays require cell fixation and are end-point assays.

The BioTracker 529 Green Pluripotent Stem Cell Dye is a live stem cell probe that selectively identifies undifferentiated human ES and iPS cells. The probe localizes at mitochondria in the human stem cells. The dye’s fluorescence is dependent on the activities of ABCB1 and ABCG2 on the cell membrane. The dye is suitable for both live cell imaging and flow cytometry applications of human pluripotent stem cells.

Spectral Properties
Absorbance: 515nm
Emission: 529nm

Application

Live cell fluorescent imaging
Research Category
Cell Imaging
Research Sub Category
Live Cell Dye

Physical form

Lyophilized

Storage and Stability

Store BioTracker 529 Green Pluripotent Stem Cell Dye at -20°C, desiccate and protect from light
Note: Centrifuge vial briefly to collect contents at bottom of vial before opening.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service