Skip to Content
Merck
All Photos(1)

Key Documents

SCR601

Sigma-Aldrich

Human Oligodendrocyte Characterization Kit

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
€626.00

€626.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 KIT
€626.00

About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.43

€626.00


Please contact Customer Service for Availability

Request a Bulk Order

species reactivity

human

Quality Level

manufacturer/tradename

Chemicon®

technique(s)

cell culture | stem cell: suitable
immunocytochemistry: suitable

input

sample type neural stem cell(s)

shipped in

dry ice

General description

EMD Millipore’s Human Oligodendrocyte Characterization Kit (SCR601) contains validated antibodies that characterize different developmental stages of oligodendrocytes along with two antibodies for the detection of neurons and astrocytes, respectively. The kit includes antibodies to NG2 Chondroitin Sulfate Proteoglycan, Myelin Proteolipid Protein, Galactocerebroside, Myelin Oligodendrocyte Glycoprotein, Glial Fibrillary Acidic Protein and MAP2. The kit provides a convenient way to characterize cell populations generated using EMD-Millipore′s Human Oligodendrocyte Differentiation Kit (SCR601)

Application

Research Category
Neuroscience

Components

Rabbit anti-NG2 Chondroitin Sulfate Proteoglycan, 10ug

Mouse anti-Myelin Proteolipid Protein, C-terminus, clone PLPC1, 20 ug

Mouse anti-Galactocerebroside, clone mGalC, 10ug

Mouse anti-Myelin Oligodendrocyte Glycoprotein (MOG), 10 ug

Mouse anti-Glial Fibrillary Acidic Protein, clone GA5, 10 ug

Mouse anti-Glial Fibrillary Acidic Protein, clone GA5, 10 ug

Storage and Stability

When stored at the recommended storage conditions (refer to Kit Components), components are stable up to the expiration date. Do not expose to elevated temperatures. Discard any remaining reagents after the expiration date.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Derivation and characterization of functional human neural stem cell derived oligodendrocyte progenitor cells (OPCs) that efficiently myelinate primary neurons in culture.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service