Skip to Content
Merck
All Photos(3)

Documents

PIHA01250

Millipore

Millicell® Standing Cell Culture Inserts

pore size 0.45 μm, diam. 12 mm, white mixed cellulose esters (MCE) membrane, size 24 wells, sterile

Synonym(s):

Millicell Cell Culture Insert, 12 mm, HA mixed cellulose esters, 0.45 µm, Millicell-HA, cell culture inserts, permeable culture insert, plate inserts, tissue culture insert, tissue culture plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104923
eCl@ss:
32011202
NACRES:
NB.22

material

polystyrene housing
white mixed cellulose esters (MCE) membrane

Quality Level

sterility

ethylene oxide treated
sterile

packaging

pack of 50

manufacturer/tradename

Millicell®

parameter

50 °C max. temp.

technique(s)

cell attachment: suitable
cell culture | mammalian: suitable
cell differentiation: suitable
toxicology assay: suitable (in vitro)

H

10.5 mm

diam.

12 mm

filtration area

0.6 cm2

size

24 wells

surface area

0.6 cm2

working volume

0.6 mL

color

opaque white, membrane

matrix

MF-Millipore

pore size

0.45 μm

application(s)

industrial qc (microbial attachment assays)

binding type

high binding surface

shipped in

ambient

General description

Device Configuration: 24-well plate, 0.45μm pore size

With Millicell® inserts, attachment or suspension cells can access media from both their apical and basolateral sides. Cell growth, structure, and function more closely mimic what occurs in vivo. In addition, Millicell® inserts make it possible to study both sides of the cell monolayer.

Millicell® Standing Inserts:
- Promotes excellent cell growth and provides an exceptional opportunity for cell studies

Membrane Type:
MF-Millipore Membrane (mixed cellulose esters)
- For exceptional anatomical and functional polarization

Applications
Cell Attachment, Cell Growth, Cell Differentiation , Immunocytochemistry

Application

Research Category
Cell Culture

Packaging

Individually blister packaged

Legal Information

MF-Millipore is a trademark of Merck KGaA, Darmstadt, Germany
Millicell is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Toluidine blue selectively stains nuclear material and acidic tissue components, aiding in histological staining for tissues rich in DNA/RNA.

Toluidine blue selectively stains nuclear material and acidic tissue components, aiding in histological staining for tissues rich in DNA/RNA.

Toluidine blue selectively stains nuclear material and acidic tissue components, aiding in histological staining for tissues rich in DNA/RNA.

Toluidine blue selectively stains nuclear material and acidic tissue components, aiding in histological staining for tissues rich in DNA/RNA.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service