Skip to Content
Merck
All Photos(3)

Documents

MTRACEBT1

Millipore

M-TRACE® Battery

for use with M-TRACE® Touch Mobile Computer

Synonym(s):

Lithium-ion Battery

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
43210000
NACRES:
JA.12

packaging

pack of -1 unit

application(s)

pharmaceutical
sterility testing

compatibility

Steritest® Symbio pump
for use with M-TRACE® Touch Mobile Computer

General description

The M-Trace® Software & Mobile App is your 21 CFR Part 11 compliant Digital QC companion for sterility testing and other regulated processes, ensuring trusted and efficient data recording. Click here to get more details on the overall M-Trace® solution.


M-TRACE® Battery is an optional accessory for M-TRACE® Touch Mobile Computer (MTRACEMC1 ) to be used as a second battery or replacement battery. This 3000 mAh Lithium-ion M-TRACE® Battery will further enable to streamline the QC sterility testing workflow.

Application

M-TRACE® Battery is an optional accessory for M-TRACE® Touch Mobile Computer that is used for sterility testing and other quality control workflows in cleanrooms and isolators.

Other Notes

Battery Type-Lithium-ion Battery
Capacity-3300mAh

Legal Information

M-TRACE is a registered trademark of Merck KGaA, Darmstadt, Germany
STERITEST is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

M-Trace® electronic test record software ensures full traceability for your sterility testing data and elevates your Steritest® Symbio pump to a whole new level. M-Trace® software reduces handling errors, ensures compliance, and boosts productivity.

M-Trace® electronic test record software ensures full traceability for your sterility testing data and elevates your Steritest® Symbio pump to a whole new level. M-Trace® software reduces handling errors, ensures compliance, and boosts productivity.

M-Trace® electronic test record software ensures full traceability for your sterility testing data and elevates your Steritest® Symbio pump to a whole new level. M-Trace® software reduces handling errors, ensures compliance, and boosts productivity.

M-Trace® electronic test record software ensures full traceability for your sterility testing data and elevates your Steritest® Symbio pump to a whole new level. M-Trace® software reduces handling errors, ensures compliance, and boosts productivity.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service