MSTSTW125
MAST® SP Sterile Tube Weld Assembly 0.125 - Sample Port
For use with MAST® Autosampling Solution SP200 Sample Pilots
Synonym(s):
STW, Sample Pilot Connection, Sterile Tube Weld
About This Item
Quality Level
storage condition
room temperature
General description
Application
Features and Benefits
- Easy to connect
- User friendly
- Single Use
Packaging
Preparation Note
Other Notes
Legal Information
related product
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Assessing on-line automated sampling as part of a PAT approach to shorten bioprocess development times through rapid, data-driven decisions.
Assessing on-line automated sampling as part of a PAT approach to shorten bioprocess development times through rapid, data-driven decisions.
Assessing on-line automated sampling as part of a PAT approach to shorten bioprocess development times through rapid, data-driven decisions.
Assessing on-line automated sampling as part of a PAT approach to shorten bioprocess development times through rapid, data-driven decisions.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service