Skip to Content
Merck
All Photos(1)

Key Documents

MABX50

Sigma-Aldrich

Anti-Chikungunya (CHIKV-Env 2)

Synonym(s):

Anti-CHIKV Antibody

Sign Into View Organizational & Contract Pricing

Select a Size

MABX50-1KC
€734.00
MABX50-10KC
€5,850.00

€734.00


Estimated to ship on18 February 2025Details


Request a Bulk Order

Select a Size

Change View
MABX50-1KC
€734.00
MABX50-10KC
€5,850.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€734.00


Estimated to ship on18 February 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

clone

monoclonal

form

solution

species reactivity

all

packaging

pkg of 1 mg (MABX50-1KC)
pkg of 10 mg (MABX50-10KC)

technique(s)

ELISA: suitable
immunodiffusion: suitable

isotype

IgG1

relevant disease(s)

infectious diseases

shipped in

wet ice

storage temp.

2-8°C

Specificity

Chikungunya Virus Env 2. No reactivity with Dengue, Zika, WNV, TBV or JEV.

Immunogen

Recombinant Chikungunya Virus Envelope 2 Protein

Application

This mouse monoclonal antibody, Cat. No. MABX50, is tested for use in Lateral Flow Assay and ELISA, for the detection of Recombinant Chikungunya Virus Envelope 2 Protein (CHIKV-E2).

Target description

Chikungunya virus is an arthropod-borne virus (arbovirus) spread by the bite of infected Aedes mosquitoes.  It is a member of the Alphavirus genus of RNA viruses.  Symptoms of fever and joint pain usually appear within a week of infection.  Muscle pain, headache, fatigue and rash may also occur.

Linkage

Recommended Matched Pair MABX50 with MABX49.

Physical form

Purified mouse monoclonal in buffer containing 0.015 M Potassium Phosphate, pH 7.2 with 0.85% Sodium Chloride and 0.05% Sodium Azide

Reconstitution

Please refer to Certificate of Analysis for concentration.

Storage and Stability

Store at 2-8 °C. For long-term storage, freeze at -20 °C

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

related product

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service