Skip to Content
Merck
All Photos(6)

Key Documents

MABT882

Sigma-Aldrich

Anti-Prodynorphin (Dynorphin B29) Antibody, clone Dyn9

clone Dyn9, from mouse

Synonym(s):

Beta-neoendorphin-dynorphin, Preprodynorphin, Preproenkephalin-B, Proenkephalin-B, Dynorphin B-29, Leumorphin

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€349.00

€349.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
€349.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

€349.00


Please contact Customer Service for Availability

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

Dyn9, monoclonal

species reactivity

human, mouse, rat

technique(s)

immunohistochemistry: suitable (paraffin)

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

rat ... Pdyn(29190)

General description

Proenkephalin-B (UniProt P06300; also known as Beta-neoendorphin-dynorphin, Preprodynorphin) is encoded by the Pdyn gene (Gene ID 29190) in rat species. Proenkephalin B is an opioid polypeptide hormone involved with chemical signal transduction and cell communication. Prodynorphin is a basic building-block of endorphins, the chemical messengers in the brain that appear most heavily involved in the anticipation and experience of pain and the formation of deep emotional bonds critical in learning and memory. Prodynorphin is produced with a signal peptide (a.a. 1-21) and a propeptide (a.a. 22-163) sequence. Following the removal of the signal and propeptide sequences, the protein is further processed by proeolytic cleavages to form Alpha-neoendorphin (a.a. 166-175), Beta-neoendorphin (a.a. 166-174), Big dynorphin (a.a. 202-233; Big Dyn), Dynorphin A(1-17) (a.a. 202-218; Dyn-A17; Dynorphin A); Dynorphin A(1-13) (a.a. 202-214), Dynorphin A(1-8) (a.a. 202-209), Leu-enkephalin (a.a. 202-206 & 221-225), Leumorphin (Dynorphin B-29) (a.a. 221-248), and Rimorphin (a.a. 221-233; Dynorphin B; Dyn-B; Dynorphin B(1-13)).

Specificity

Clone Dyn9 recognizes preproenkephalin-B (a.a. 1-248 of rat sequence), proenkephalin-B (a.a. 22-248 of rat sequence), and leumorphin/dynorphin B-29 (a.a. 221-248 of rat sequence), but not other peptides derived from Proenkephalin-B.

Immunogen

Thyroglobulin-conjugatged linear peptide corresponding to rat leumorphin (dynorphin B-29) C-terminal half sequence.

Application

Immunohistochemistry Analysis: A 1:20 dilution from a representative lot detected Prodynorphin/Dynorphin B29 immunoreactivity in human midbrain tissue sections.

Immunohistochemistry Analysis: A 1:20 dilution of clone Dyn9 hybridoma culture supernatant detected Prodynorphin/Dynorphin B29 immunoreactivity in human (hippocampus and cerebral cortex), mouse (fundus striati), and rat (nucleus supraopticus, lateral hipothalamic nucleus, paraventricular nucleus, and nucleus accumbens, septum pellucidum, caudate-putamen, and hippocampus) brain tissue sections (Courtesy of Ricardo Martínez Murillo, PhD, Cajal Institute, Madrid Spain).
This mouse monoclonal Anti-Prodynorphin (Dynorphin B29), clone Dyn9 Antibody, Cat. No. MABT882 is validated for use in Immunohistochemistry (Paraffin) for the detection of prodynorphin and leumorphin/dynorphin B-29.

Quality

Evaluated by Immunohistochemistry in human thalamus tissue sections.

Immunohistochemistry Analysis: A 1:20 dilution of this antibody detected Prodynorphin/Dynorphin B29 immunoreactivity in human thalamus tissue sections.

Target description

28.08/28.06/28.39 kDa (Rat/Mouse/Human preproenkephalin-B), 25.85/25.76/26.28 kDa (Rat/Mouse/Human proenkephalin-B), 3.320/3.320/3.352 kDa (Rat/Mouse/Human leumorphin) calculated.

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phillip R Kramer et al.
Journal of pain research, 17, 2311-2324 (2024-07-08)
Herpes Zoster in humans is the result of varicella zoster virus (VZV) infection. Injecting rats with varicella zoster virus produces pain similar to herpes zoster "shingles" pain in humans. . In a previous study, orofacial pain was induced by injecting

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service