Skip to Content
Merck
All Photos(1)

Key Documents

MABT528-AF488

Sigma-Aldrich

Anti-Connexin 43 Antibody, CT Antibody, clone 1C5.1 Antibody, Alexa Fluor 488 Conjugate

clone 1C5.1, from mouse, ALEXA FLUOR 488

Synonym(s):

Gap junction alpha-1 protein, Cx43, Gap junction 43 kDa heart protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43
Pricing and availability is not currently available.

biological source

mouse

Quality Level

conjugate

ALEXA FLUOR 488

antibody form

purified antibody

antibody product type

primary antibodies

clone

1C5.1, monoclonal

species reactivity

human, rat

technique(s)

immunocytochemistry: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... GJA1(2697)

General description

Gap junction alpha-1 protein (UniProt: P17302; also known as Connexin-43, Cx43, Gap junction 43 kDa heart protein) is encoded by the GJA1 (also known as GJAL) gene (gene ID: 2697) in human. Cx43 is a member of the connexin family of proteins and a component of gap junctions. Connexins belong to a family of integral membrane proteins that assemble as hexamers (connexons) to form gap junctions, which are specialized sites of cell-cell contact. Gap junctions allow the passage of ions, metabolites and low molecular weight messenger molecules to pass from one cell to the other. Cx43 undergoes rapid turnover in the cell and its monomers may reside in the ER/Golgi network, forming a reservoir available for assembly upon degradation of existing connexin-43 channels. Cx43 may play a critical role in the physiology of hearing by participating in the recycling of potassium to the cochlear endolymph. It also acts as a negative regulator of bladder functional capacity by enhancing intercellular electrical and chemical transmission, thus sensitizing bladder muscles to cholinergic stimuli and causing them to contract. Cx43 is reported to be essential for normal cardiac development. Mice lacking Cx43 die soon after birth due to cardiac malformations and obstruction of the pulmonary artery. Cx43 has been detected in a variety of tissues including: atrial and ventricular myocytes, granulose cells, astrocytes, fibroblasts, leptomeningeal cells, brain, lung, smooth muscle, uterine muscle, kidney, lens, epithelium, cornea, and testis. Cx43 is phosphorylated at Ser368 by PKC gamma; which leads to disassembly of gap junction plaques and inhibition of gap junction activity. Mutations in GJA1 gene are reported to cause Oculodentodigital dysplasia and Hallermann-Streiff syndrome.

Specificity

Anti-Connexin 43, CT antibody, clone 1C5.1, Alexa Fluor 488 Conjugate specificaly detects Connexin 43 in human and rat. This antibody has been shown to bind to an epitope between amino acids 315-382.

Immunogen

GST/His-tagged recombinant fragment corresponding to 68 amino acids from the C-terminal half of human Connexin 43.

Application

The unconjugated antibody (Cat. No. MABT528) is shown to be suitable also for Flow Cytometry, Immunohistochemistry, Immunofluorescence, and Immunoprecipitation applications.
This mouse monoclonal Anti-Connexin 43 Antibody, CT, clone 1C5.1, Alexa Fluor 488 Conjugate, Cat. No. MABT528-AF488 is used for the detection of Connexin 43 by Immunocytochemistry.

Quality

Evaluated by Immunocytochemistry in HeLa cells.

Immunocytochemistry Analysis: A 1:100 dilution of this antibody detected Connexin 43 in HeLa cells.

Target description

43.00 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Other Notes

Concentration: Please refer to lot specific datasheet.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service