Skip to Content
Merck
All Photos(1)

Documents

MABT153

Sigma-Aldrich

Anti-Aggrecan Antibody (full length), clone 4D7.1

clone 4D7.1, 1 mg/mL, from mouse

Synonym(s):

Aggrecan core protein, Cartilage-specific proteoglycan core protein, CSPCP, Chondroitin sulfate proteoglycan core protein 1, Chondroitin sulfate proteoglycan 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

4D7.1, monoclonal

species reactivity

human

concentration

1 mg/mL

technique(s)

western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ACAN(176)

General description

Aggrecan is an extracellular protein that is also known as Aggrecan core protein, Cartilage-specific proteoglycan core protein (CSPCP), Chondroitin sulfate proteoglycan core protein 1, and Chondroitin sulfate proteoglycan 1. Aggrecan is encoded by the gene ACAN/AGC1/CSPG1/MSK16. Aggrecan is a proteoglycan that is a major component of the extracellular matrix of cartilaginous tissues. The major function of Aggrecan is to resist compression in cartilage, thus providing protection for joint movement and against injury. Aggrecan binds various other ECM proteins via its large globular domains. Aggrecan can be cleaved into Aggrecan core protein 2, a C-terminal fragment component that decreases in cartilage as people age. Aggrecan is expressed in cartilage and is a massive (>2500 kDa) proteoglycan protein that has multiple isoforms. The appearance of Aggrecan fragments in the blood is a marker for various joint diseases.

Immunogen

KLH-conjugated linear peptide corresponding to human Aggrecan.

Application

Research Category
Cell Structure
Research Sub Category
ECM Proteins
This Anti-Aggrecan Antibody (full length), clone 4D7.1 is validated for use in Western Blotting for the detection of Aggrecan (full length).

Quality

Evaluated by Western Blotting in human chondrocyte cell lysate.

Western Blotting Analysis: 2 µg/mL of this antibody detected Aggrecan (full length) in 10 µg of human chondrocyte cell lysate.

Target description

~270 kDa observed. The calculated molecular weight is 250 kDa However, this protein is known to be highly glycosylated and may be observed at higher molecular weights.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service