Skip to Content
Merck
All Photos(2)

Key Documents

MABS1916

Sigma-Aldrich

Anti-c-Met Antibody, clone seeMet 13

clone seeMet 13, from mouse

Synonym(s):

Hepatocyte growth factor receptor, HGF receptor, HGF/SF receptor, Proto-oncogene c-Met, Scatter factor receptor, SF receptor, Tyrosine-protein kinase Met

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

seeMet 13, monoclonal

species reactivity

human

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunoprecipitation (IP): suitable
inhibition assay: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... HGF(3082)

Related Categories

General description

Hepatocyte growth factor receptor (EC 2.7.10.1; UniProt P08581; also known as HGF receptor, HGF/SF receptor, Proto-oncogene c-Met, Scatter factor receptor, SF receptor, Tyrosine-protein kinase Met) is encoded by the MET (also known as AUTS9, HCC, RCCP) gene (Gene ID 4233) in human. HGF receptor or c-Met is initially produced as a 1390-a.a. precursor protein with an N-terminal signal peptide, postranslational proteolytic cleavage yields the mature tyrosine kinase receptor composed of an extracellular alpha chain (a.a. 25-307) linked via a disulfide bond to a transmembrane beta chain with an extracellular domain (a.a. 308-932), transmembrane helix (a.a. 933-955), and a cytoplasmic domain (a.a. 956-1390). The alpha-chain and the extracellular portion of beta-chain form the Sema domain that mediates receptor dimerization and ligand binding. The cytoplasmic portion of beta-chain contains the kinase domain (a.a. 1078-1345) and a C-terminal tail that contains a docking region (a.a. 1212-1390) for effector and adapter proteins essential for downstream signaling. Hepatocyte growth factor (HGF) is the only known c-Met ligand. HGF binding induces c-Met dimerization and autophosphorylation on Y1230, Y1234 and Y1235, leading to a conformation change and further phosphorylation on Y1349 and Y1356, making C-terminal tail docking region available for adaptor and signalling molecules. c-Met mediates a wide-range of biological activities, including cell proliferation, motility, angiogenesis and morphogenesis, and is an attractive target for cancer therapy. Aberrant c-Met activation/signalling promotes cancer cell proliferation and is associated with poor prognosis in various human cancers. Upregulated c-Met expression is also known to cause resistance against HER2, EGFR and B-RAF inhibitory drugs in cancer treatment.

Specificity

Clone seeMet 13 bound strongly to native c-Met by flow cytometry, while exhibiting low reactivity and specificity towards denatured c-Met by Western blotting (Wong, J.S., et al. (2013). Oncotarget. 4(7):1019-1036).

Immunogen

His-tagged recombinant human c-Met alpha chain (Wong, J.S., et al. (2013). Oncotarget. 4(7):1019-1036).

Application

Detect Hepatocyte growth factor receptor using this mouse monoclonal Anti-c-Met, clone seeMet 13, Cat. No. MABS1916, validated for use in Flow Cytometry, Immunocytochemistry, Immunoprecipitation, and Inhibition studies.
Flow Cytometry Analysis: 0.2 µL from a representative lot detected c-Met immunoreactivity on the surface of one million SNU-5 cells.

Flow Cytometry Analysis: A representative lot immunostained live SNU-5 cells. A slight decrease in antibody immunoreactivity was observed when the temperature was dropped from 37°C to 4°C (Wong, J.S., et al. (2013). Oncotarget. 4(7):1019-1036).

Immunocytochemistry Analysis: A representative lot immunostained live SNU-5 cells. Indirect fluorescence labelling following subsequent cell fixation and permeabilization revealed increased antibody cytoplasmic internalization when antibody incubation was performed at 37°C than at 4°C (Wong, J.S., et al. (2013). Oncotarget. 4(7):1019-1036).

Immunoprecipitation Analysis: A representative lot immunoprecipitated c-Met from SNU-5 cell lysate (Wong, J.S., et al. (2013). Oncotarget. 4(7):1019-1036).

Inhibition Analysis: A representative lot prevented HGF-induced scatter of HaCaT cells. Clone seeMet 13 inhibited cell proliferation by blocking cell division without inducing apoptosis (Wong, J.S., et al. (2013). Oncotarget. 4(7):1019-1036).

Note: seeMet 13 exhibited low reactivity and specificity towards denatured c-Met by Western blotting. This monoclonal antibody is not recommended for Western blotting application (Wong, J.S., et al. (2013). Oncotarget. 4(7):1019-1036).
Research Category
Signaling

Quality

Evaluated by Immunocytochemistry in SNU-5 cells.

Immunocytochemistry Analysis: A 1:500 dilution of this antibody detected both surface and internalized c-Met by fluorescent immunocytochemistry staining of 4% paraformaldehyde-fixed, 0.3% Triton X-100-permeabilized SNU-5 cells.

Target description

32.51 kDa (alpha chain; a.a. 25-307), 155.5/153.0 kDa (pro-/mature uncleaved isoform 1), 157.7/155.2 kDa (pro-/mature uncleaved isoform 2), 85.75/83.25 kDa (pro-/mature uncleaved isoform 3) calculated.

Physical form

Format: Purified
Protein G purified.
Purified mouse IgG1 in PBS without preservatives.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.

Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service