Skip to Content
Merck
All Photos(2)

Documents

MABN2609

Sigma-Aldrich

Anti-P2X4 receptor Antibody, clone 12-10H

Synonym(s):

P2X purinoceptor 4;P2X4;ATP receptor;Purinergic receptor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

12-10H, monoclonal

mol wt

calculated mol wt 43.5 kDa

purified by

using protein G

species reactivity

rat

packaging

antibody small pack of 100 μg

technique(s)

ELISA: suitable
dot blot: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable

isotype

IgG1κ

epitope sequence

N-terminal half

Protein ID accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

General description

P2X purinoceptor 4 (UniProt: P51577; also known as P2X4, ATP receptor, Purinergic receptor) is encoded by the P2rx4 gene (Gene ID: 29659) in rat. P2 receptors comprise P2X ionotropic receptors and G protein-coupled P2Y receptors. P2X receptors are expressed in many tissues, where they are involved in a number of functions including synaptic transmission, muscle contraction, platelet aggregation, inflammation, macrophage activation, neuropathic and inflammatory pain. P2X4, one of the most sensitive purinergic receptors, is a multi-pass membrane glycoprotein with two cytoplasmic domains (1-33 and 360-388), two transmembrane domains (aa 34-54 and 339-359), and one extracellular domain (aa 55-338). It is widely distributed in the brain and its strong expression is observed in microglial cells. P2X4 serves as a receptor for extracellularly released ATP and acts as a ligand-gated ion channel. It is shown to be preferentially localized in lysosomes, where it is protected from proteolysis by its glycosylation state. At low pH, high ATP levels in the lysosomes do not activate P2X4. However, it is activated by intra-lysosomal ATP only in its fully dissociated tetra-anionic form when the pH increases to 7.4. Hence, P2X4 functions as a Ca2+-channel following the fusion of late endosomes and lysosomes. P2X4 is reported to participates in basal T-cell activity without TCR/CD3 stimulation. It also promotes the differentiation and activation of Th17 cells via expression of retinoic acid-related orphan receptor C/RORC. P2X4 also contributes to regulation of synaptic strength through participation in the formation of long-term potentiation. Microglial P2X4 receptors are shown to be upregulated during nerve-injury associated neuropathic pain. Clone 12-10H recognize the native structure of head domain of P2X4. (Ref.: Igawa, T., et al (2019). Purinergic Signalling 15(1);27-35; Suurvälia, J., et al. (2017). Biomed. J. 40(5); 245-256; Tsuda, M., et al. (2003). Nature. 424(6950); 778-783).

Specificity

Clone 12-10H is a mouse monoclonal antibody that detects P2X purinoreceptor 4. It targets an epitope within the N-terminal half.

Immunogen

A KLH-conjugated recombinant fragment derived from the head domain of P2X4 receptor expressed in E.coli.

Application

Quality Control Testing

Evaluated by Immunohistochemistry (Paraffin) in rat cerebral cortex tissue sections.

Immunohistochemistry (Paraffin) Analysis (IHC(P)): A 1:50 dilution of this antibody detected P2X4 in rat cerebral cortex tissue sections.

Tested applications

ELISA Analysis: A representative lot detected P2X4 receptor in ELISA applications (Igawa, T., et al. (2019). Purinergic Signal.15(1):27-35).

Dot Blot: A representative lot detected P2X4 receptor in Dot Blot applications (Igawa, T., et al. (2019). Purinergic Signal. 15(1):27-35).

Immunocytochemistry Analysis: A representative lot detected P2X4 receptor in Dot Blot applications (Igawa, T., et al. (2019). Purinergic Signal. 15(1):27-35).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-P2X4 receptor, clone 12-10H, Cat. No. MABN2609, is a mouse monoclonal antibody that detects P2X purinoceptor 4 and is tested for use in DOT Blot, ELISA, Immunocytochemistry, and Immunohistochemistry (Paraffin).

Physical form

Purified mouse monoclonal antibody IgG1 in PBS without preservatives.

Storage and Stability

Store at -10°C to -25°C. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service