Skip to Content
Merck
All Photos(1)

Documents

MABN2299

Sigma-Aldrich

Anti-L1CAM/CD171 Antibody, clone S10.33

Synonym(s):

CD171, N-CAM-L1, NCAM-L1, Neural cell adhesion molecule L1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rat

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

S10.33, monoclonal

species reactivity

mouse

packaging

antibody small pack of 100

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable

isotype

IgG2aκ

epitope sequence

Extracellular domain

UniProt accession no.

Gene Information

mouse ... L1cam(16728)

Specificity

Clone S10.33 is a rat monoclonal antibody that detects L1CAM/CD171. It targets an epitope within the extracellular domain.

Immunogen

Recombinant fragment corresponding to the entire extracellular domain of mouse L1CAM/CD171.

Application

Quality Control Testing

Evaluated by Immunocytochemistry in Neuro2A cells.

Immunocytochemistry Analysis: A 1:100 dilution of this antibody detected L1CAM/CD171 in Neuro2A cells.

Tested Applications

Flow Cytometry Analysis: A representative lot detected L1CAM/CD171 in Flow Cytometry applications (Maddaluno, L., et al. (2009). J Exp Med. 206(3):623-35).

Immunohistochemistry Applications: A representative lot detected L1CAM/CD171 in Immunohistochemistry applications (Maddaluno, L., et al. (2009). J Exp Med. 206(3):623-35).

Immunofluorescence Analysis: A representative lot detected L1CAM/CD171 in Immunofluorescence applications (Maddaluno, L., et al. (2009). J Exp Med. 206(3):623-35; Magrini, E., et al. (2014). J Clin Invest. 124(10):4335-50).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Neural cell adhesion molecule L1 (UniProt: P11627; also known as N-CAM-L1, NCAM-L1, CD171) is encoded by the L1cam (also known as Caml 1) gene (Gene ID: 16728) in murine species. L1CAM is a single-pass type I membrane glycoprotein that is synthesized with a signal peptide (aa 1-19), which is subsequently cleaved off to produce the mature form that contains an extracellular domain (aa 20-1123), a transmembrane domain (aa 1124-1146), and a cytoplasmic domain (aa 1147-1260). The large extracellular domain contains six Ig-like C2 type domains and five fibronectin type III repeats. It serves as a cell recognition molecule within the nervous system that plays a critical role in multiple processes, including neuronal migration, axonal growth and fasciculation, and synaptogenesis during brain development. In the mature brain, it plays a role in the dynamics of neuronal structure and function, including synaptic plasticity. Its expression is also observed in tumor vasculature. Tumor-bearing mice that lack L1CAM exhibit reduced angiogenesis and improved vascular stabilization that leads to reduced tumor growth and metastasis. These effects have been attributed to L1CAM-induced IL-6-mediated STAT3 phosphorylation and inhibition of IL-6/JAK/STAT signaling axis that prevents endothelial cell proliferation and migration. (Ref.: Magrini, A., et al. (2014). J. Clin. Invest. 124(10); 4335-4350; Maddaluno, L., et al. (2009). J. Exp. Med. Vol. 206(3); 623-635).

Physical form

Purified rat monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Reconstitution

1.0 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service