Skip to Content
Merck
All Photos(1)

Documents

MABN1840

Sigma-Aldrich

Anti-Neuritin (CPG15) Antibody, clone A6.1

clone A6.1, from mouse

Synonym(s):

candidate plasticity gene 15, CPG15

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

A6.1, monoclonal

species reactivity

rat, human

species reactivity (predicted by homology)

monkey (based on 100% sequence homology), bovine (based on 100% sequence homology)

packaging

antibody small pack of 25 μg

technique(s)

western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... NRN1(51299)

General description

Neuritin (UniProt: Q9NPD7) is encoded by the NRN1 (also known as NRN) gene (Gene ID: 51299) in human. Neuritin, a conserved, glycosylphosphatidylinositol (GPI)-anchored protein that plays a role in the development, survival and function of neurons. Its activity is induced by brain-derived neurotrophic factor (BDNF) and neurotrophin-3 (NT-3). It is highly expressed by induced and natural regulatory T cells (TREGs). It also promotes the maintenance of the TREGs pool by stabilizing Foxp3 expression and promoting the persistence of these cells. Deficiency of Neuritin is shown to result in an imbalance between central and peripheral TREG populations and functions. Neuritin has a wide variety of effects in the nervous system, such as promoting neurite outgrowth, modulating neurite outgrowth during neuronal differentiation, and protecting motor neuron axons. It can also promote dendritic growth, regulate synaptic plasticity, and stabilize active synapses, and promote synaptic maturation. Its expression in the hippocampus is shown to be induced by neuronal activity following chemical- or electrical-induced seizures, ischemia, and exercise. Chronic antidepressant treatment is reported to increase neuritin expression in rat brain. Neuritin is synthesized with a signal peptide (aa 1-27) and a propeptide (aa 28-116) that are cleaved off in the mature form (aa 117-142). (Ref.: Son, H., et al. (2012). Proc. Natl. Acad. Sci. USA. 109(28); 11378-83; Zhou, S., and Zhou, J (2014). Curr. Med. Chem. 21(10); 1212-19). Note: A6.1 hybridoma clone, and WT/KO validation, courtesy of Lauren Makuch and Dr. Elly Nedivi, Massachusetts Institute of Technology.

Specificity

Clone A6.1 detects human and rat Neuritin.

Immunogen

GST-tagged full-length human recombinant Neuritin.

Application

Anti-Neuritin (CPG15), clone A6.1, Cat. No. MABN1840, is a highly specific mouse monoclonal antibody that targets Neuritin and has been tested for use in Western Blotting.
Research Category
Neuroscience

Quality

Evaluated by Western Blotting with recombinant human/rat neuritin.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected recombinant human/rat Neuritin (CPG15).

Target description

~15 kDa observed; 15.33 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service