Skip to Content
Merck
All Photos(1)

Key Documents

MABN1785

Sigma-Aldrich

Anti-CD166 (ALCAM) Antibody, clone TAG-1A3

clone TAG-1A3, from mouse

Synonym(s):

Activated leukocyte cell adhesion molecule, CD166 antigen

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
€153.00
200 μL
€375.00

€153.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
€153.00
200 μL
€375.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€153.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

TAG-1A3, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μL

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunofluorescence: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... ALCAM(214)

Related Categories

General description

CD166 antigen (UniProt: Q13740; also known as Activated leukocyte cell adhesion molecule, CD166) is encoded by the ALCAM (also known as MEMD) gene (Gene ID: 214) in human. CD166 is a cell adhesion molecule that mediates both heterotypic cell-cell contacts via its interaction with CD6 and homotypic cell-cell contacts. It also promotes T-cell activation and proliferation via its interactions with CD6 and is shown to be involved in the formation and maturation of the immunological synapse via its interactions with CD6. CD166 mediates attachment of dendritic cells onto endothelial cells via homotypic interaction and inhibits endothelial cell migration and promotes endothelial tube formation via homotypic interactions. It has an extracellular (aa 28-527), transmembrane (aa 528-549), and a cytoplasmic (aa 550-583) domain. CD166 is expressed by activated T-cells, B-cells, monocytes and thymic epithelial cells. It is also detected on hematopoietic stem cells derived from umbilical cord blood, on lymph vessel endothelial cells, skin and tonsil, and on peripheral blood monocytes. Lower levels of CD166 are detected in spleen, placenta, and liver. CD166 is also detected in highly metastasizing melanoma cell lines.

Specificity

Clone TAG-1A3 specifically detects CD166 in human.

Immunogen

Corneal endothelial cells (hCENC) isolated from donated human cadaves and immunized with monophosphoryl-lipid A trehalose dicorynomycolate (MPLCTDM) adjuvant.

Application

Anti-CD166 (ALCAM) Antibody, clone TAG-1A3, Cat. No. MABN1785, is a highly specific mouse monoclonal antibody that targets CD166 and has been tested in Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, and Western Blotting.
Flow Cytometry Analysis: A representative lot detected CD166 (ALCAM) in Flow Cytometry applications (Ding, Y., et. al. (2014). MAbs. 6(6):1439-52).

Immunocytochemistry Analysis: A representative lot detected CD166 (ALCAM) in Immunocytochmistry applications (Ding, Y., et. al. (2014). MAbs. 6(6):1439-52).

Immunoprecipitation Analysis: A representative lot detected CD166 (ALCAM) in Immunoprecipitation applications (Ding, Y., et. al. (2014). MAbs. 6(6):1439-52).

Flow Cytometry Analysis: A represetative lot detected CD166 (ALCAM) in human embryonic stem cells, human corneal stromal fibroblast (hCSF) cells, immortalized human corneal endothelial cells (B4G12), and human endothelial cells from donors (2) (Courtesy of Dr. Vanessa Ding, Ph.D. and Dr. Andre Choo, Ph.D, BTI, Singapore).

Immunofluorescence Analysis: A representative lot detected CD166 (ALCAM) in Immunofluorescence applications (Ding, Y., et. al. (2014). MAbs. 6(6):1439-52).

Western Blotting Analysis: A representative lot detected CD166 (ALCAM) in Western Blotting applications (Ding, Y., et. al. (2014). MAbs. 6(6):1439-52).
Research Category
Neuroscience

Quality

Identity Confirmation by Isotyping Test.

Isotyping Analysis: The identity of this monoclonal antibody is confirmed by isotyping test to be human IgG1 .

Target description

~80 kDa observed; 65.10 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Meng Feng et al.
Frontiers in cell and developmental biology, 9, 716610-716610 (2021-10-15)
Hedgehog (HH) signaling plays a critical role in osteoarthritis (OA) pathogenesis, but the molecular mechanism remains to be elucidated. We show here that Sonic Hedgehog (SHH) gene expression is initiated in human normal cartilage stromal cells (NCSC) and increased in
Mahmoud Yousefifard et al.
Journal of stem cells & regenerative medicine, 18(2), 53-63 (2023-01-31)
Few studies are conducted on the efficacy of human adipose-derived stem cells (ADSCs) in spinal cord injury (SCI) management and electrophysiological changes in the spinal cord. Therefore, the present study aimed to determine the effect of ADSCs on neuropathic pain

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service