Skip to Content
Merck
All Photos(3)

Documents

MABF854

Sigma-Aldrich

Anti-S100A9 Antibody, clone 6B4

clone 6B4, from mouse

Synonym(s):

Protein S100-A9, Calgranulin-B, Calprotectin L1H subunit, Leukocyte L1 complex heavy chain, Migration inhibitory factor-related protein 14, MRP-14, p14, S100 calcium-binding protein A9, S100A9

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6B4, monoclonal

species reactivity

human

should not react with

mouse

technique(s)

ELISA: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... S100A9(6280)

General description

Protein S100-A9 (UniProt P06702; also known as Calgranulin-B, Calprotectin L1H subunit, Leukocyte L1 complex heavy chain, Migration inhibitory factor-related protein 14, MRP-14, p14, S100 calcium-binding protein A9, S100A9) is encoded by the S100A9 (also known as 60B8AG, CAGB, CFAG, CGLB, L1AG, LIAG, MAC387, MRP14) gene (Gene ID 6280) in human. S100A9 is one of the 21 members of the S100 family of proteins. S100A8 (calgranulin-A), S100A9 (calgranulin-B) and the heterdimer S100A8/S100A9 (calprotectin) exert their immunological and inflammatory action via binding to TLR-4, RAGE and some other putative receptors. S100A8, calprotectin, and S100A9 are chemoattractants for neutrophils, and S100A9 is known to induce the expression and activation of beta2-integrins. S100A9 is also shown to stimulate neutrophil degranulation and phagocytosis, while both S100A8 and S100A9 can induce the secretion of inflammatory cytokines from human monocytes. In addition, S100A9 is shown to potentiate both fMLP- and GM-CSF-stimulated IL-8 production from neutrophils, although S100A9 on its own cannot induce neutrophil cytokine production in the absence of fMLP and GM-CSF.

Specificity

Clone 6B4 selectively detects human S100A9 with no cross-reactivity toward hS100A8, hS100A12, mS100A8, or mS100A9.

Immunogen

Recombinant protein corresponding to human S100A9.

Application

Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology
This mouse monoclonal Anti-S100A9 Antibody, clone 6B4, Cat. No. MABF854 is validated for use in Western Blotting and ELISA, for the detection of S100A9.
Western Blotting Analysis: A representative lot detected recombinant hS100A9 (rhS100A9), as well as endogenous hS100A9 in neutrophil extract and purified calprotectin (S100A8/S100A9 dimer), but not rhS100A8 & rhS100A12 or recombinant murine S100A8 & S100A9 (Dr Philippe Tessier, Laval University, Quebec, CA).
ELISA Analysis: A representative lot detected recombinant human S100A9 (rhS100A9) by direct/non-sandwich ELISA, but not rhS100A8 & rhS100A12 or recombinant murine S100A8 & S100A9 (Dr Philippe Tessier, Laval University, Quebec, CA).
ELISA Analysis: A representative lot detected recombinant human S100A9 (rhS100A9) as well as endogenous S100A9 in purified human calprotectin (S100A8/S100A9 dimer) by sandwich ELISA, but not rhS100A8 or rhS100A12 (Dr Philippe Tessier, Laval University, Quebec, CA).

Quality

Evaluated by Western Blotting in human spleen tissue lysate.

Western Blotting Analysis: 0.1 µg/mL of this antibody detected S100A9 in 10 µg of human spleen tissue lysate.

Target description

~13 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ antibody in buffer containing PBS without preservatives.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lauren R Begg et al.
Communications medicine, 4(1), 22-22 (2024-02-21)
Understanding why some triple-negative breast cancer (TNBC) patients respond poorly to existing therapies while others respond well remains a challenge. This study aims to understand the potential underlying mechanisms distinguishing early-stage TNBC tumors that respond to clinical intervention from non-responders

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service