Skip to Content
Merck
All Photos(2)

Key Documents

MABF3177

Sigma-Aldrich

Anti-M-CSF Antibody, clone 3G6.4.8

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies, primary antibodies

clone

3G6.4.8, monoclonal

mol wt

calculated mol wt 60.18 kDa
observed mol wt ~37 kDa

purified by

using protein G

species reactivity

human

packaging

antibody small pack of 100 μg

technique(s)

immunofluorescence: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG1κ

epitope sequence

Unknown

Protein ID accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... CSF1(1435)

General description

Macrophage colony-stimulating factor 1 (UniProt: P09603; also known as CSF-1, M-CSF, MCSF, Lanimostim) is encoded by the CSF1 gene (Gene ID: 1435) in human. M-CSF is a single-pass type I membrane glycoprotein that is synthesized with a signal peptide (aa 1-32), which is subsequently cleaved off to generate the mature form that contains a lumenal domain (aa 33-496), a transmembrane domain (aa 497-517), and a cytoplasmic domain (aa 518-554). It can be present as a disulfide-linked homodimer or heterodimer. Two distinct biologically active forms of M-CSF have been described: a membrane-bound and a secreted form that result from alternative splicing of the CSF1 gene. It plays an essential role in the regulation of survival, proliferation, and differentiation of hematopoietic precursor cells, especially mononuclear phagocytes, such as macrophages and monocytes. It promotes the release of proinflammatory chemokines, and thereby plays an important role in innate immunity and in inflammatory processes. M-CSF is also involved in the regulation of osteoclast proliferation and differentiation, the regulation of bone resorption, and for normal bone development. Its biological effects are mediated via binding to CSF-1R/c-Fms receptor that is expressed by multipotent hematopoietic cells, mononuclear phagocyte progenitor cells, monocytes, tissue macrophages, and osteoclasts. Aberrant expression of M-CSF or CSF-1R is reported to promote cancer cell proliferation, invasion and formation of metastases. Overexpression of M-CSF or CSF-1R is observed in a significant percentage of breast, ovarian, prostate, and endometrial cancers.

Specificity

Clone 3G6.4.8 is a mouse monoclonal antibody that detects human macrophage colony-stimulating factor 1 (M-CSF). It targets an epitope within the N-terminal half.

Immunogen

A recombinant fragment corresponding to 149 amino acids from the N-terminal half of human Macrophage colony-stimulating factor 1 (M-CSF) with a GHERQSEGS sequence attached at its C-terminal end.

Application

Quality Control Testing

Evaluated by Western Blotting with recombinant Human M-CSF.

Western Blotting Analysis (WB): A 1:100,000 dilution of this antibody detected recombinant human M-CSF under non-denaturing conditions.

Tested Applications


Immunohistochemistry (Paraffin) Analysis: A 1:50 dilution from a representative lot detected M-CSF in human tonsil tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-M-CSF, clone 3G6.4.8, Cat. No. MABF3177, is a mouse monoclonal antibody that detects M-CSF and is tested for use in Immunohistochemistry (Paraffin) and Western Blotting.

Physical form

Purified mouse monoclonal antibody IgG1 in PBS with 0.05% sodium azide.

Storage and Stability

Recommend storage at +2°C to +8°C. For long term storage antibodies can be kept at -20°C. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service