Skip to Content
Merck
All Photos(1)

Documents

MABF2073

Sigma-Aldrich

Anti-Rabies Virus CVS-11 Antibody, clone 509-6

clone 509-6, from mouse

Synonym(s):

Glycoprotein, Glycoprotein Rabies virus, Rabies virus strain CVS-11

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

509-6, monoclonal

species reactivity

rabies virus

species reactivity (predicted by homology)

all

packaging

antibody small pack of 25 μg

technique(s)

ELISA: suitable
flow cytometry: suitable
neutralization: suitable

isotype

IgG2aκ

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

General description

Glycoprotein (UniProt: O92284) is encoded by the G gene in Rabies virus, strain CVS-11. Glycoprotein is a single-pass type I membrane protein that belongs to the lyssavirus glycoprotein family. It attached the virus to host cellular receptor, inducing endocytosis of the virion. In the endosome, the acidic pH induces conformational changes in the glycoprotein trimer, which trigger fusion between virus and cell membrane. The muscular form of the nicotinic acetylcholine receptor (nAChR), the neuronal cell adhesion molecule (NCAM), and the p75 neurotrophin receptor (p75NTR) are reported to bind glycoprotein and thereby facilitate rabies virus entry into cells. Glycoprotein structure includes a virion surface (aa 20-459), helical region (aa 460-480) and an intravirion (aa 481-524). Arginine 352 is reported to be involved in Rabies virus pathogenicity and its mutation attenuates the virus. Glycoprotein is glycosylated and palmitoylated by host and glycosylation is considered to be crucial for glycoprotein export at the cell surface. Clone 509-6 is shown to neutralize all fixed and street rabies virus strains except some virus isolates from bats. The conformational antigenic site I for this clone is located at aa 231. (Ref.: Wiktor, TJ and Koprowski, H (1980). J. Exp. Med. 152: 99-112; Marissen, WE et al (2005). J. Virol. 79(8): 4672 4678).

Specificity

Clone 509-6 detected glycoprotein from Rabies virus, strain CVS-11 in L929 cells expressing the rabies protein. It did not detect glycoprotein in wild type L929 cells.

Immunogen

Beta-propiolactone inactivated Rabies virus, strain CVS-11 (RABV).

Application

Anti-Rabies Virus, clone 509-6, Cat. No. MABF2073, is a mouse monoclonal antibody that detects glycoprotein of Rabies virus, strain CVS-11 and has been tested for use in ELISA, Flow Cytometry, Fluorescence Activated Cell Sorting (FACS), and Neutralizing.
Neutralizing Analysis: A representative lot neutralized Rabies Virus (Schumacher, C.L., et. al. (1989). J Clin Invest. 84(3):971-5; Bunschoten, H., et. al. (1989). J Gen. Virol. 70 ( Pt 2):291-8; Marissen, W.E., et. al. (2005). J Virol. 79(8):4672-8).

ELISA Analysis: A representative lot detected Rabies Virus in ELISA applications (Marissen, W.E., et. al. (2005). J Virol. 79(8):4672-8).

Fluorescence Activated Cell Sorting (FACS) Analysis: A representative lot detected Rabies Virus in Fluorescence Activated Cell Sorting (FACS) applications (Marissen, W.E., et. al. (2005). J Virol. 79(8):4672-8).
Research Category
Inflammation & Immunology

Quality

Evaluated by Flow Cytometry in L929 cells expressing the rabies protein and in L929 (WT) cells.

Flow Cytometry Analysis: 1 µg of this antibody detected glycoprotein of Rabies virus in one million L929 cells expressing the rabies protein.

Target description

58.86 kDa calculated.

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG2a in PBS without azide.

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service