Skip to Content
Merck
All Photos(2)

Key Documents

MABE918

Sigma-Aldrich

Anti-Brd4 Antibody, clone 1F11

clone 1F11, from mouse

Synonym(s):

Bromodomain-containing protein 4, Protein HUNK1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€379.00

€379.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
€379.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€379.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1F11, monoclonal

species reactivity

human

technique(s)

ChIP: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... BRD4(23476)

General description

Bromodomain-containing protein 4 (Brd4), or Mitotic chromosome-associated protein (MCAP), is a chromatin reader protein and kinase that recognizes and binds acetylated histones and plays a key role in the transmission of epigenetic memory across cell divisions and transcription regulation. Brd4 remains associated with acetylated chromatin throughout the entire cell cycle and provides epigenetic memory for post mitotic G1 gene transcription by preserving acetylated chromatin status and maintaining high-order chromatin structure. During interphase, Brd4 plays a key role in regulating the transcription of signal-inducible genes by associating with the P-TEFb complex and recruiting it to promoters. Brd4 is widely expressed and localized to the nucleus and acetylated chromatin. Brd4 also plays a large role in many cancers, and through its regulation of transcription, Brd4 is important in multiple cellular processes including DNA damage responses, aging, and development.

Immunogen

Epitope: a.a. 785-1055 domain
Recombinant protein corresponding to the a.a. 785-1055 domain of human Brd4.

Application

Anti-Brd4, clone 1F11 is an antibody against Brd4 for use in WB, ChIP.
Chromatin Immunoprecipitation Analysis: A representative lot immunoprecipitated Brd4 using breast cancer cells MCF-7 (Data courtesy of Cheng-Ming Chiang, Simmons Comprehensive Cancer Center, University of Texas Southwestern Medical Center, Dallas, Texas).

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected Brd4 in 16 µg of HeLa cell lysate.

Target description

~152 kDa observed. Uncharacterized band(s) may appear in some lysates.

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service