Skip to Content
Merck
All Photos(1)

Documents

MABC1759

Sigma-Aldrich

Anti-CD48 Antibody, clone 1B4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1B4, monoclonal

mol wt

calculated mol wt 27.68 kDa

purified by

using protein G

species reactivity

human

packaging

antibody small pack of 100 μg

technique(s)

flow cytometry: suitable
inhibition assay: suitable

isotype

IgG2a

epitope sequence

Extracellular domain

Protein ID accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... CD48(962)

General description

CD48 antigen (UniProt: P09326; also known as B-lymphocyte activation marker, BLAST-1, BCM1 surface antigen, Leukocyte antigen MEM-102, SLAM family member 2, SLAMF2, Signaling lymphocytic activation molecule 2, TCT.1, CD48) is encoded by the CD48 (also known as BCM1, BLAST1) gene (Gene ID: 962) in human. CD48 is synthesized with a signal peptide (aa 1-26) and a propeptide (aa 221-243) that are subsequently cleaved off to produce the mature form (aa 27-220). CD48 exists in both a membrane-bound and soluble form. It is anchored to the cell membrane by a GPI linkage that allows it to associate with cholesterol-rich lipid rafts and associated signaling molecules. It is expressed in mature lymphocytes and monocytes, but not on non-hematopoietic tissues. It serves as a ligand for CD2 and facilitates interaction between activated lymphocytes. It participates in adhesion and activation of immune cells and has activating role on T cells, antigen presenting cells, and granulocytes. CD48 expression is shown to increase under inflammatory conditions and upon exposure to interferon , β, and . Bacterial and viral infections can also increase its expression on human peripheral blood mononuclear cells. CD48 is reported to be constitutively expressed at high levels on almost all multiple myeloma (MM) plasma cells. Clone 1B4 is shown to induce significant cytotoxic effect against MM cells in vitro and in vivo. (Ref.: McArdel, SL., et al. (2016). Clin. Immunol. 164; 10-20; Hosen, N., et al. (2011). Br. J. Haematol. 156(2); 213-224;).

Specificity

Clone 1B4 is a mouse monoclonal antibody that detects human CD48. It targets an epitope within the extracellular domain.

Immunogen

BaF3 cells expressing human CD48.

Application

Quality Control Testing

Evaluated by Flow Cytometry in Raji cells.

Flow Cytometry Analysis: 1 μg of this antibody detected CD48 in one million Raji cells.

Tested Applications

Flow Cytometry Analysis: A representative lot detected CD48 in Flow Cytometry applications (Hosen, N., et al. (2012). Br J Haematol. 156(2):213-24).

Inhibition Assay: A representative lot of this antibody inhibited tumor growth in severe combined immunodeficient mice inoculated subcutaneously with multiple myeloma cells. (Hosen, N., et al. (2012). Br J Haematol. 156(2):213-24).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-CD48, clone 1B4, Cat. No. MABC1759, is a mouse monoclonal antibody that detects CD48 and is tested for use in Flow Cytometry and Inhibition Assays.

Physical form

Purified mouse monoclonal antibody IgG2a in PBS without azide.

Storage and Stability

Store at -10°C to -25°C. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service