MAB8906B
Anti-Measles Antibody, nucleoprotein, clone 83KKII, biotin-conjugated
clone 83KKII, Chemicon®, from mouse
Synonym(s):
Rubeola
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
biological source
mouse
Quality Level
conjugate
biotin conjugate
antibody form
purified antibody
antibody product type
primary antibodies
clone
83KKII, monoclonal
species reactivity
human
manufacturer/tradename
Chemicon®
technique(s)
ELISA: suitable
isotype
IgG
shipped in
wet ice
Specificity
MAB8906B recognizes the measles nucleoprotein by capture EIA.
Immunogen
Epitope: nucleoprotein
Application
Capture EIA: 1:5,000-1:10,000.
Final working dilutions must be determined by end user.
Final working dilutions must be determined by end user.
Research Category
Infectious Diseases
Infectious Diseases
Research Sub Category
Infectious Diseases - Viral
Infectious Diseases - Viral
This Anti-Measles Antibody, nucleoprotein, clone 83KKII, biotin-conjugated is validated for use in ELISA for the detection of Measles.
Physical form
Purified by Protein-A Sepharose™. 1.0 mg/mL in PBS.
Storage and Stability
Maintain at -20°C in convenient aliquots for up to 12 months.
Legal Information
CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
Sepharose is a trademark of Cytiva
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
The journal of gene medicine, 14(9-10), 590-597 (2012-09-28)
We aimed to determine the feasibility of monitoring viral delivery and initial distribution to solid tumors using iodinated contrast agent and micro-computed tomography (CT). Human BxPC-3 pancreatic tumor xenografts were established in nude mice. An oncolytic measles virus with an
Quantitative molecular imaging of viral therapy for pancreatic cancer using an engineered measles virus expressing the sodium-iodide symporter reporter gene.
Ajr. American Journal of Roentgenology null
Gene therapy, 19(3), 279-287 (2011-07-15)
The purpose of our study was to validate the ability of pinhole micro-single-photon emission computed tomography/computed tomography (SPECT/CT) to: 1) accurately resolve the intratumoral dispersion pattern and 2) quantify the infection percentage in solid tumors of an oncolytic measles virus
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service