Skip to Content
Merck
All Photos(1)

Key Documents

MAB3706

Sigma-Aldrich

Anti-CD4 Antibody, intracellular, clone 024-10D6.B3

clone 024-10D6.B3, Chemicon®, from mouse

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€381.00

€381.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
€381.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€381.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

clone

024-10D6.B3, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
electrophoretic mobility shift assay: suitable
immunocytochemistry: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Specificity

Recognizes the cytoplasmic tail of CD4. Many functions have been assigned to this tail, including anti-viral regulation (down/up regulation of CD4), T-cell maturation, activation and differentiation, and Lck signal transduction, among others. This antibody is useful as a control for studies involving targeting of intracellular protein domains of T cells.

Immunogen

Synthetic peptide corresponding to amino acids 439-458 of the cytoplasmic tail of human CD4.

Application

Detect CD4 using this Anti-CD4 Antibody, intracellular, clone 024-10D6.B3 validated for use in ELISA, EMSA, FUNC & IC.
Immunocytochemistry

ELISA

Stimulation/Inhibition of biological activity

Electrophoretic mobility shift analysis

Optimal working dilutions must be determined by end user.

Physical form

Format: Purified
Purified immunoglobulin. Liquid in 0.02 M phosphate buffer, 0.25M NaCl, with 0.1% sodium azide.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service