Recommended Products
biological source
mouse
Quality Level
conjugate
unconjugated
antibody form
purified antibody
antibody product type
primary antibodies
clone
3E5, monoclonal
mol wt
calculated mol wt 198.04 kDa
observed mol wt ~200 kDa
purified by
using protein G
species reactivity
rat, human
packaging
antibody small pack of 100 μL
technique(s)
ELISA: suitable
electron microscopy: suitable
western blot: suitable
isotype
IgG
epitope sequence
Unknown
Protein ID accession no.
UniProt accession no.
shipped in
dry ice
target post-translational modification
unmodified
Gene Information
human ... lamb1> LAMB1(3912)
General description
Specificity
Immunogen
Application
Evaluated by Western Blotting in Human placenta tissue lysates.
Western Blotting Analysis: A 1:500 dilution of this antibody detected Laminin β1 in Human placenta tissue lysates.
Tested Applications
Western Blotting Analysis: A representative lot detected Laminin β1 in Western Blotting applications (Engvall, E., et al. (1986). J Cell Biol.;103(6 Pt1):2457-65).
Electron Microscopy: A representative lot detected Laminin β1 in Electron Microscopy applications (Engvall, E., et al. (1986). J Cell Biol.;103(6 Pt1):2457-65).
Inhibition: A representative lot inhibited the neurite-promoting activity of laminin. (Engvall, E., et al. (1986). J Cell Biol.;103(6 Pt1):2457-65).
ELISA Analysis: A representative lot detected Laminin β1 in ELISA applications (Engvall, E., et al. (1986). J Cell Biol.;103(6 Pt1):2457-65).
Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Physical form
Storage and Stability
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service