Skip to Content
Merck
All Photos(1)

Key Documents

KAA002FR

Sigma-Aldrich

Cellular Senescence Assay Kit

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
€290.00

€290.00


Please contact Customer Service for Availability


Select a Size

Change View
1 KIT
€290.00

About This Item

UNSPSC Code:
41116158
NACRES:
NA.84

€290.00


Please contact Customer Service for Availability

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 3 - Repr. 1B

Storage Class Code

3 - Flammable liquids

Flash Point(F)

135.5 °F

Flash Point(C)

57.5 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service