HX0628
Hydrofluoric Acid
OmniTrace Ultra
Synonym(s):
Hydrogen Fluoride
About This Item
Recommended Products
grade
for inorganic trace analysis
Quality Level
Assay
47-51%
form
liquid
concentration
≥50-70% in hydrofluoric acid
pH
(at 20 °C (68 °F)strongly acid)
bp
106 °C/1013 hPa
mp
-35 °C
solubility
soluble (Water: Soluble)
density
1.16 g/cm3 at 20 °C (68 °F)
shipped in
ambient
storage temp.
room temp
SMILES string
F
InChI
InChI=1S/FH/h1H
InChI key
KRHYYFGTRYWZRS-UHFFFAOYSA-N
Application
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Eye Dam. 1 - Skin Corr. 1A
Storage Class Code
6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service