Skip to Content
Merck
All Photos(1)

Key Documents

HTS156M

Sigma-Aldrich

ChemiSCREEN Membrane Preparation Recombinant Human MC 1 Melanocortin Receptor

Sign Into View Organizational & Contract Pricing

Select a Size

200 UNITS
€688.00

€688.00


Please contact Customer Service for Availability


Select a Size

Change View
200 UNITS
€688.00

About This Item

UNSPSC Code:
41106514
eCl@ss:
32161000
NACRES:
NA.45

€688.00


Please contact Customer Service for Availability

biological source

human

Quality Level

recombinant

expressed in Chem-1 cells

form

liquid

manufacturer/tradename

ChemiScreen
Chemicon®

technique(s)

ligand binding assay: suitable (GTPγS)
radioligand binding assay (RLBA): suitable

NCBI accession no.

UniProt accession no.

General description

The melanocortin-1 receptor (MC1) is a member of the G-protein-coupled receptor family of proteins that is responsible for regulating skin and hair color. The MC1 receptor is regulated by the physiological agonists alpha-melanocyte-stimulating hormone (alphaMSH) and adrenocorticotropic hormone (ACTH), and antagonist agouti signaling protein (ASP) (Rouzud et al., 2005). In 1995 a landmark study demonstrated that over 80% of humans with red hair or fair skin have a dysfunctional variant of the MC1 gene (Valverde et al., 1995). MC1 is, therefore, a major determinant of sun sensitivity and genetic risk factor for melanoma and non-melanoma skin cancer. Millipore′s MC1 membrane preparations are crude membrane preparations made from our proprietary stable recombinant cell lines to ensure high-level of GPCR surface expression; thus, they are ideal HTS tools for screening of agonists and antagonists of MC1. The membrane preparations exhibit a Kd of 1.3nM for [125I]-[Nle4, D-Phe7]-α-MSH (NDP-αMSH). With 0.3 nM [125I]-NDP-αMSH, 5μg/well MC1 Membrane Prep typically yields greater than 50-fold signal-to-background ratio.

Application

Human MC1 GPCR membrane preparation for Radioligand binding Assays & GTPγS binding.
Radioligand binding assay and GTPγS binding.

Biochem/physiol Actions

  • GPCR Class: A
  • Protein Target: MC1
  • Target Sub-Family: Melanocortin

Physical form

Liquid in packaging buffer: 50 mM Tris pH 7.4, 10% glycerol and 1% BSA with no preservatives. Packaging method: Membranes protein were adjusted to 1 mg/mL in packaging buffer, and dispensed at 1 mL/vial. Vials were rapidly frozen, and stored at -80°C.

Storage and Stability

Maintain frozen at -70°C for up to 2 years. Do not freeze and thaw.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service