Skip to Content
Merck
All Photos(1)

Key Documents

HTS050M

Sigma-Aldrich

ChemiScreen H1 Membrane Preparation

Human H1 GPCR membrane preparation for Radioligand binding Assays & GTPγS binding.

Sign Into View Organizational & Contract Pricing

Select a Size

200 UNITS
€688.00

€688.00


Please contact Customer Service for Availability


Select a Size

Change View
200 UNITS
€688.00

About This Item

UNSPSC Code:
41106514
eCl@ss:
32161000
NACRES:
NA.84

€688.00


Please contact Customer Service for Availability

biological source

human

Quality Level

recombinant

expressed in Chem-3 cells

packaging

pkg of 2 mL

manufacturer/tradename

ChemiScreen
Chemicon®

concentration

0.5 mg/mL

technique(s)

ligand binding assay: suitable (GTPγS)
radioligand binding assay (RLBA): suitable

UniProt accession no.

shipped in

dry ice

General description

Full-length human HRH1 cDNA
Histamine exerts its biological effects through a family of four GPCRs. Stimulation of the H1 histamine receptor causes smooth muscle contraction, induces vascular permeability, and stimulates catecholamine release from the adrenal medulla. H1 is also widely expressed in brain, and activation of H1 excites several types of neurons by blocking potassium conductance (Hill et al., 1997).

Chemicon′s H1 membrane preparations are crude membrane preparations made from our proprietary stable recombinant cell lines to ensure high-level of GPCR surface expression; thus, they are ideal HTS tools for screening of antagonists of H1 interactions and its ligands. The membrane preparations exhibit a Kd of 4.5 nM for [3H]-Pyrilamine. With 3 nM [3H]-Pyrilamine, 5 mg/well and 10 mg/well H1 Membrane Prep typically yield greater than 3 and 6-fold signal-to-background ratios, respectively.

Application

Radioligand binding assay, and GTPγS binding.

Biochem/physiol Actions

GPCR Class: A
Protein Target: H1
Target Sub-Family: Histamine

Quality

Signal:background and specific binding values obtained in a competition binding assay with varying amounts of TP membrane prep:
10 µg/well
Signal:Background 6.6
Specific Binding (cpm) 925



SPECIFICATIONS: 1 unit = 10 µg for most lots. Specific activity (units/µg) may vary between lots.
Bmax: 8.3 pmol/mg
Kd: 4.5 nM
Note: Lot number MM1357015 contains 2 mL at 2 mg/mL, for which 1 unit = 20 µg

Specifications

Inucbation Conditions
Membranes are mixed with radioactive ligand and unlabeled competitor (see Figures 1 and 2 for concentrations tested) in binding buffer in a nonbinding 96-well plate, and incubated for 1-2 h. Prior to filtration, a GF/C 96-well filter plate is coated with 0.33% polyethyleneimine for 30 min, then washed with 50 mM HEPES, pH 7.4, 0.5% BSA. Binding reaction is transferred to the filter plate, and washed 3 times (1 mL per well per wash) with Wash Buffer. The plate is dried and counted.

Binding buffer: 50 mM Tris, pH 7.4, 10 mM MgCl2, 1 mM EDTA, filtered and stored at 4°C

Radioligand: [3H] Pyrilamine (Perkin Elmer#:NET594250UC )

Wash Buffer: 50 mM Tris, pH 7.4 filtered and stored at 4°C.

One package contains enough membranes for at least 200 assays (units), where an unit is the amount of membrane that will yield greater than 3-fold signal:background with 3H-labeled Pyrilamine at 3 nM

Physical form

Liquid in packaging buffer: 50 mM Tris pH 7.4, 10% glycerol and 1% BSA no preservatives. Packaging method: Membranes protein were adjusted to 0.5 mg/mL in 1 mL packaging buffer, rapidly frozen, and stored at -80°C.

Storage and Stability

Maintain frozen at -70°C for up to 2 years. Do not freeze and thaw.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service