Skip to Content
Merck
All Photos(1)

Key Documents

HAWG03700

Millipore

MCE Membrane Filter, 0.45 μm Pore Size

greener alternative

MF-Millipore, filter diam. 37 mm, hydrophilic, white, gridded

Synonym(s):

MF-Millipore Membrane Filter, 0.45 µm pore size, gridded

Sign Into View Organizational & Contract Pricing

Select a Size

100 EA
€244.00

€244.00


Please contact Customer Service for Availability


Select a Size

Change View
100 EA
€244.00

About This Item

UNSPSC Code:
40161507
eCl@ss:
32031602
NACRES:
NB.24

€244.00


Please contact Customer Service for Availability

material

cellulose membrane
gridded
mixed cellulose esters (MCE) membrane
white filter

Quality Level

Agency

in accordance with OSHA ID 160

description

37 mm diameter, mixed cellulose esters (MCE) membrane, hydrophilic, white, gridded, 100 discs

sterility

non-sterile

feature

hydrophilic

manufacturer/tradename

MF-Millipore
Millipore

sustainability

Greener Alternative Product

parameter

4 L/min-cm2 air flow rate
60 mL/min-cm2 water flow rate
75 °C max. temp.

diam.

37 mm

filter diam.

37 mm

thickness

150 μm

color

white

refractive index

n/D 1.51

matrix

MF-Millipore

pore size

0.45 μm pore size
79 % porosity

capacity

155 μg/cm2 adsorption capacity (insulin)

bubble point

≥1.8 bar, air with water at 23 °C

greener alternative category

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

General description

Biologically inert mixtures of cellulose acetate and cellulose nitrate have made MF-Millipore membrane filters one of the most widely used membranes in analytical and research applications.

MF-Millipore filters without Triton surfactant contain minimum amounts of wetting agent and have a lower water extractable content than standard MF-Millipore filters.

Features & Benefits:
- Versatile filter for biological and environmental monitoring applications
- Available in a range of pore sizes, colored black or white, with or without a gridded surface
- Compatible with ethylene oxide, gamma irradiation, and autoclave sterilization methods
Filter Code: HAWG
We are committed to bringing you Greener Alternative Products, which have been re-engineered to enable more sustainable research solutions. The inner & outer boxes containing the membrane discs are made from bio-circular material allocated using the ISCC mass balance approach. Click here for more information.

Application

  • Clarification of aqueous solutions
  • Particle removal
  • Particle analysis
  • Microbiology analysis
  • Isolation of virus-like particles in wastewater
  • Microplastics analysis grade water
  • Nucleic acid binding, including eDNA

Other Notes

Directions for Use

  • Organism Retention: Microorganism
  • Mode of Action: Filtration (size exclusion)
  • Application: General laboratory filtration
  • Intended Use: Retention or removal of biological contaminants
  • Instructions for Use: Sterilizing filtration of a liquid through a membrane with a 0.2 μm (or smaller) pore size effectively removes biological contaminants, including bacteria, mold and yeast. For the selective retention of larger biological contaminents, liquid filtration through membranes with 0.45 μm (or larger) pore sizes may be used to trap and support microorganism growth for subsequent culture and analysis
  • Storage Statement: Store in dry location away from heat source
  • Disposal Statement: Dispose of in accordance with applicable federal, state and local regulations.

Legal Information

MF-Millipore is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cheng-Der Liu et al.
Journal of virology, 94(7) (2020-01-17)
The binding of Epstein-Barr Virus (EBV) nuclear antigen 1 (EBNA1) to the latent replication origin (oriP) triggers multiple downstream events to support virus-induced pathogenesis and tumorigenesis. Although EBV is widely recognized as a B-lymphotropic infectious agent, little is known about

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service