Key Documents
GT20/25
Millipore
NovAseptic® Mixer
Multi Tool for Mixing Head and Male Bearing, GMP and USM
About This Item
UNSPSC Code:
41103801
eCl@ss:
32010600
material
aluminum body
stainless steel key (handle)
Quality Level
product line
NovAseptic®
weight
1.2 kg (2.65 lb)
Application
A dynamometric wrench is not included but can be supplied, order on Catalogue No. GT/24-5. The Multi Tool is not recommended for use on larger sizes of mixing heads than GMP2000. Mixing heads GMP5000 - GMP20000 shall be lifted by hand, for tightening the male bearing choose tool GT30/26.
Other Notes
Surface Finish, Exterior: Anodized aluminium, not on key handle.
Legal Information
NOVASEPTIC is a registered trademark of Merck KGaA, Darmstadt, Germany