Skip to Content
Merck
All Photos(1)

Key Documents

FCCH025111

Sigma-Aldrich

FlowCellect Oxidative Stress Characterization Kit

Oxidative stress has been found to play a key role in a number of pathological disorders.

Synonym(s):

FlowCellect

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000

species reactivity (predicted by homology)

all

manufacturer/tradename

FlowCellect

technique(s)

flow cytometry: suitable

compatibility

for use with EasyCyte 8HT
for use with EasyCyte Mini
for use with EasyCyte Plus
for use with EasyCyte

detection method

fluorometric

shipped in

dry ice

General description

Oxidative stress has been found to play a key role in a number of pathological disorders. To aid researchers studying this phenomenon Millipore has developed the FlowCellect Oxidative Stress Characterization Kit. This kit contains the chemical and immunological reagents necessary to detect carbonyl groups introduced onto proteins by reactive oxygen species (ROS). The test method involves derivatization of protein carbonyl groups with 2,4-dinitrophenylhydrazine (DNPH). This chemical reaction results in proteins being covalently coupled to DNP at their carbonyl sites. The DNP-derivatized proteins are then incubated with a FITC conjugated monoclonal antibody that specifically binds to the DNP moiety. Subsequent detection with a bench-top Guava EasyCyte Plus flow cytometer allows quantification of the extent of oxidative stress within cells. Although the assay and all of the kit components were optimized using a Guava instrument, any flow cytometer can be utilized provided it contains a blue laser (488 nm) light source.

Application

Research Category
Neuroscience

Components

Anti-DNP -FITC

Derivatization Control Solution

5X Assay Buffer

DNPH Solution

10X Wash Buffer

User Manual

Storage and Stability

Upon arrival the FITC conjugated anti-DNP antibody should be transferred to a -20°C freezer. The remaining kit components (FCCH025111-1) should be stored at 2 - 8ºC.
All kit components are stable for up to six (6) months from date of receipt if stored and handled appropriately.

Legal Information

FlowCellect is a trademark of Luminex Corporation

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Corrosion

Signal Word

Warning

Precautionary Statements

Storage Class Code

8B - Non-combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cristina Talarico et al.
Oncotarget, 7(13), 15868-15884 (2016-02-26)
Glioblastoma multiforme (GBM) is the most aggressive CNS tumor and is characterized by a very high frequency of clinical relapse after therapy and thus by a dismal prognosis, which strongly compromises patients survival. We have recently identified the small molecule

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service